Product Image
Thermo Scientific™

Potassium Chloride Solution

Potassium Chloride Solution (pH 2.2, 0.2M) is used as a pretreatment to increase yield of Legionella from clinical specimens.

Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidad
R06262120 x 4,5 mL
R062620100 x 4,5 mL
Número de catálogo R062621
Precio (EUR)
-
Cantidad:
20 x 4,5 mL

Pretreat the samples to increase yield of Legionella from clinical specimens using Thermo Scientific™ Remel™ Potassium Chloride Solution (pH 2.2, 0.2M). In 1981, Bopp et al. demonstrated the recovery rate of Legionella from environmental water samples was improved by treating samples with acid prior to plating onto selective media1. Buesching et al. demonstrated the recovery of Legionella organisms from clinical specimens was improved by pre-treatment with potassium chloride-hydrochloric acid solution (pH 2.2)2.

Potassium Chloride Solution (pH 2.2, 0.2M) is a liquid medium that is used as a pretreatment to increase yield of Legionella from clinical specimens. Potassium Chloride Solution (pH 2.2, 0.2M) is an acid wash treatment which serves to decontaminate specimens prior to inoculation of media for Legionella cultivation.

Potassium Chloride Solution (pH 2.2, 0.2M) contains hydrochloric acid (0.2 M) and potassium chloride (0.2M) with pH 2.2.

  1. Bopp, C.A., J.W. Summer, G.K. Morris, and J.G. Wells. 1981. J. Clin. Microbiol. 13:714-719.
  2. Buesching, W.J., R.A. Brust, and L.W. Ayers. 1983. J. Clin. Microbiol. 17:1153-1155.
Especificaciones
DescripciónPotassium Chloride Solution
Formato15mm x 103mm Tube
Cantidad20 x 4,5 mL
Tipo de productoBuffer Solution
Unit SizeEach