Orange DNA Loading Dye (6X)
Thermo Scientific™

Orange DNA Loading Dye (6X)

Thermo Scientific 6X Orange Loading Dye is used to prepare DNA markers and samples for loading on agarose or polyacrylamideRead more
Have Questions?
Catalog NumberQuantity
R06315 x 1 mL
Catalog number R0631
Price (USD)
48.65
Online exclusive
58.25
Save 9.60
Each
Quantity:
5 x 1 mL
Request bulk or custom format
Price (USD)
48.65
Online exclusive
58.25
Save 9.60
Each
Thermo Scientific 6X Orange Loading Dye is used to prepare DNA markers and samples for loading on agarose or polyacrylamide gels. It contains two different dyes (xylene cyanol FF and orange G) for visual tracking of DNA migration during electrophoresis. The presence of glycerol ensures that the DNA in the ladder and sample forms a layer at the bottom of the well. The EDTA included in the solutions binds divalent metal ions and inhibits metal-dependent nucleases.

6X Orange DNA Loading Dye is used for conventional DNA electrophoresis.

Highlights

• Two-color tracking of DNA migration during DNA electrophoresis
• No DNA masking during gel exposure to UV light
• EDTA binds divalent metal ions and inhibits metal dependent nucleases

Applications

• Analysis of small DNA molecules
• Preparation of DNA ladders, markers, and samples for loading on agarose or polyacrylamide gels
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Gel CompatibilityAcrylamide Gels, Agarose Gels
Label or DyeOrange G, Xylene Cyanol FF (XCFF)
Product TypeDNA Loading Dye
Quantity5 x 1 mL
Concentration6X
Unit SizeEach

Frequently asked questions (FAQs)

What can I use as a substitution for the discontinued RapidRun Loading Dye?

We recommend using the Orange DNA Loading Dy (6X) (Cat. No. R0631) as a substitute for the RapidRun Loading Dye.