Search Thermo Fisher Scientific
Thermo Scientific™ Remel Schaedler Broth is for the cultivation of fastidious aerobic and anaerobic microorganisms.
Format | 15mm x 103mm Tube |
Type | Broth |
Description | Schaedler Broth |
Product Type | Broth |
Quantity | 100 x 5 mL |
Unit Size | Each |
Format | 15mm x 103mm Tube |
Type | Broth |
Description | Schaedler Broth |
Product Type | Broth |
Quantity | 20 x 5 mL |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Quantity | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
R064480 | Full specifications | Each | 100 x 5 mL | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R064482 | Full specifications | Each | 20 x 5 mL | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Thermo Scientific™ Remel Schaedler Broth is for the cultivation of fastidious aerobic and anaerobic microorganisms.