Thermo Scientific™
Tryptic Soy Broth
Catalog number: R064900
Thermo Scientific™
Tryptic Soy Broth
Catalog number: R064900
Cultivate and isolate microorganisms with ease using Thermo Scientific™ Tryptic Soy Broth, a general purpose enrichment broth.
Catalog Number
R064900
Unit Size
Each
Format
15mm x 103mm Tube
Fill Volume
4 mL
Quantity
100/Pk.
Price (USD)
Full specifications
DescriptionTryptic Soy Broth
Product TypeBroth
Manufacturing OriginUS
Quantity100/Pk.
Certifications/ComplianceIndustrial Reference: AOAC, BAM, COMPF, SMWW, IDF, SMD, USDA, USP.
Fill Volume4 mL
TypeBroth
Format15mm x 103mm Tube
Unit SizeEach
Catalog Number
Unit Size
Each
Format
15mm x 103mm Tube
Fill Volume
4 mL
Quantity
20/Pk.
Price (USD)
Full specifications
DescriptionTryptic Soy Broth
Product TypeBroth
Manufacturing OriginUS
Quantity20/Pk.
Certifications/ComplianceIndustrial Reference: AOAC, BAM, COMPF, SMWW, IDF, SMD, USDA, USP.
Fill Volume4 mL
TypeBroth
Format15mm x 103mm Tube
Unit SizeEach
Catalog Number
Unit Size
Each
Format
15mm x 103mm Tube
Fill Volume
4.5 mL
Quantity
100/Pk.
Price (USD)
Full specifications
DescriptionTryptic Soy Broth
Product TypeBroth
Manufacturing OriginUS
Quantity100/Pk.
Certifications/ComplianceIndustrial Reference: AOAC, BAM, COMPF, SMWW, IDF, SMD, USDA, USP.
Fill Volume4.5 mL
TypeBroth
Format15mm x 103mm Tube
Unit SizeEach
Catalog Number
Unit Size
Each
Format
15mm x 103mm Tube
Fill Volume
4.5 mL
Quantity
20/Pk.
Price (USD)
Full specifications
DescriptionTryptic Soy Broth
Product TypeBroth
Manufacturing OriginUS
Quantity20/Pk.
Certifications/ComplianceIndustrial Reference: AOAC, BAM, COMPF, SMWW, IDF, SMD, USDA, USP.
Fill Volume4.5 mL
TypeBroth
Format15mm x 103mm Tube
Unit SizeEach
Catalog Number
Unit Size
Each
Format
Tube
Fill Volume
5 mL
Quantity
100/Pk.
Price (USD)
Full specifications
ColorClear liquid, light to medium amber
DescriptionTryptic Soy Broth
Product TypeBroth
Manufacturing OriginUS
Quantity100/Pk.
Certifications/ComplianceAOAC, BAM, COMPF, SMWW, IDF, SMD, USDA, USP
Fill Volume5 mL
Packaging TypeGlass tube with black screw cap
TypeBroth
FormatTube
Unit SizeEach
Catalog Number
Unit Size
Each
Format
Tube
Fill Volume
10 mL
Quantity
100/Pk.
Price (USD)
Full specifications
ColorClear liquid, light to medium amber
DescriptionTryptic Soy Broth
Product TypeBroth
Manufacturing OriginUS
Quantity100/Pk.
Certifications/ComplianceAOAC, BAM, COMPF, SMWW, IDF, SMD, USDA, USP
Fill Volume10 mL
Packaging TypeGlass tube with black screw cap
TypeBroth
FormatTube
Unit SizeEach
Catalog Number
Unit Size
Each
Format
20mm x 113mm Tube
Fill Volume
10 mL
Quantity
100/Pk.
Price (USD)
Full specifications
DescriptionTryptic Soy Broth
Product TypeBroth
Manufacturing OriginUS
Quantity100/Pk.
Certifications/ComplianceIndustrial Reference: AOAC, BAM, COMPF, SMWW, IDF, SMD, USDA, USP.
Fill Volume10 mL
TypeBroth
Format20mm x 113mm Tube
Unit SizeEach
Catalog Number
Unit Size
Each
Format
20mm x 113mm Tube
Fill Volume
10 mL
Quantity
20/Pk.
Price (USD)
Full specifications
DescriptionTryptic Soy Broth
Product TypeBroth
Manufacturing OriginUS
Quantity20/Pk.
Certifications/ComplianceIndustrial Reference: AOAC, BAM, COMPF, SMWW, IDF, SMD, USDA, USP.
Fill Volume10 mL
TypeBroth
Format20mm x 113mm Tube
Unit SizeEach
Catalog Number
Unit Size
Each
Format
20mm x 113mm Tube
Fill Volume
18 mL
Quantity
20/Pk.
Price (USD)
Full specifications
DescriptionTryptic Soy Broth
Product TypeBroth
Manufacturing OriginUS
Quantity20/Pk.
Certifications/ComplianceIndustrial Reference: AOAC, BAM, COMPF, SMWW, IDF, SMD, USDA, USP.
Fill Volume18 mL
TypeBroth
Format20mm x 113mm Tube
Unit SizeEach
Catalog Number
Unit Size
Each
Format
16mm x 125mm Tube
Fill Volume
9 mL
Quantity
100/Pk.
Price (USD)
Full specifications
DescriptionTryptic Soy Broth
Product TypeBroth
Manufacturing OriginUS
Quantity100/Pk.
Certifications/ComplianceIndustrial Reference: AOAC, BAM, COMPF, SMWW, IDF, SMD, USDA, USP.
Fill Volume9 mL
TypeBroth
Format16mm x 125mm Tube
Unit SizeEach
Showing 10 of 20Show more
Catalog NumberSpecificationsUnit SizeFormatFill VolumeQuantityPrice (USD)
R064900Full specifications
Each15mm x 103mm Tube4 mL100/Pk.Request A Quote
DescriptionTryptic Soy Broth
Product TypeBroth
Manufacturing OriginUS
Quantity100/Pk.
Certifications/ComplianceIndustrial Reference: AOAC, BAM, COMPF, SMWW, IDF, SMD, USDA, USP.
Fill Volume4 mL
TypeBroth
Format15mm x 103mm Tube
Unit SizeEach
R064902Full specifications
Each15mm x 103mm Tube4 mL20/Pk.Request A Quote
DescriptionTryptic Soy Broth
Product TypeBroth
Manufacturing OriginUS
Quantity20/Pk.
Certifications/ComplianceIndustrial Reference: AOAC, BAM, COMPF, SMWW, IDF, SMD, USDA, USP.
Fill Volume4 mL
TypeBroth
Format15mm x 103mm Tube
Unit SizeEach
R065000Full specifications
Each15mm x 103mm Tube4.5 mL100/Pk.Request A Quote
DescriptionTryptic Soy Broth
Product TypeBroth
Manufacturing OriginUS
Quantity100/Pk.
Certifications/ComplianceIndustrial Reference: AOAC, BAM, COMPF, SMWW, IDF, SMD, USDA, USP.
Fill Volume4.5 mL
TypeBroth
Format15mm x 103mm Tube
Unit SizeEach
R065002Full specifications
Each15mm x 103mm Tube4.5 mL20/Pk.Request A Quote
DescriptionTryptic Soy Broth
Product TypeBroth
Manufacturing OriginUS
Quantity20/Pk.
Certifications/ComplianceIndustrial Reference: AOAC, BAM, COMPF, SMWW, IDF, SMD, USDA, USP.
Fill Volume4.5 mL
TypeBroth
Format15mm x 103mm Tube
Unit SizeEach
R07222Full specifications
EachTube5 mL100/Pk.Request A Quote
ColorClear liquid, light to medium amber
DescriptionTryptic Soy Broth
Product TypeBroth
Manufacturing OriginUS
Quantity100/Pk.
Certifications/ComplianceAOAC, BAM, COMPF, SMWW, IDF, SMD, USDA, USP
Fill Volume5 mL
Packaging TypeGlass tube with black screw cap
TypeBroth
FormatTube
Unit SizeEach
R07226Full specifications
EachTube10 mL100/Pk.Request A Quote
ColorClear liquid, light to medium amber
DescriptionTryptic Soy Broth
Product TypeBroth
Manufacturing OriginUS
Quantity100/Pk.
Certifications/ComplianceAOAC, BAM, COMPF, SMWW, IDF, SMD, USDA, USP
Fill Volume10 mL
Packaging TypeGlass tube with black screw cap
TypeBroth
FormatTube
Unit SizeEach
R08942Full specifications
Each20mm x 113mm Tube10 mL100/Pk.Request A Quote
DescriptionTryptic Soy Broth
Product TypeBroth
Manufacturing OriginUS
Quantity100/Pk.
Certifications/ComplianceIndustrial Reference: AOAC, BAM, COMPF, SMWW, IDF, SMD, USDA, USP.
Fill Volume10 mL
TypeBroth
Format20mm x 113mm Tube
Unit SizeEach
R08944Full specifications
Each20mm x 113mm Tube10 mL20/Pk.Request A Quote
DescriptionTryptic Soy Broth
Product TypeBroth
Manufacturing OriginUS
Quantity20/Pk.
Certifications/ComplianceIndustrial Reference: AOAC, BAM, COMPF, SMWW, IDF, SMD, USDA, USP.
Fill Volume10 mL
TypeBroth
Format20mm x 113mm Tube
Unit SizeEach
R08948Full specifications
Each20mm x 113mm Tube18 mL20/Pk.Request A Quote
DescriptionTryptic Soy Broth
Product TypeBroth
Manufacturing OriginUS
Quantity20/Pk.
Certifications/ComplianceIndustrial Reference: AOAC, BAM, COMPF, SMWW, IDF, SMD, USDA, USP.
Fill Volume18 mL
TypeBroth
Format20mm x 113mm Tube
Unit SizeEach
R117832Full specifications
Each16mm x 125mm Tube9 mL100/Pk.Request A Quote
DescriptionTryptic Soy Broth
Product TypeBroth
Manufacturing OriginUS
Quantity100/Pk.
Certifications/ComplianceIndustrial Reference: AOAC, BAM, COMPF, SMWW, IDF, SMD, USDA, USP.
Fill Volume9 mL
TypeBroth
Format16mm x 125mm Tube
Unit SizeEach
Showing 10 of 20Show more
Remove the need for pipetting, dispensing or autoclaving with convenient, pre-filled and ready-to-use TSB tubes.
Part numbers beginning with R11 are for sterility testing according to USP (United States Pharmacopeia).
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.