Remel™ Butterfield's with 0.1% Peptone Phosphate Buffer
Product Image
Thermo Scientific™

Remel™ Butterfield's with 0.1% Peptone Phosphate Buffer

Thermo Scientific™ Remel Butterfield's with 0.1% Peptone Phosphate Buffer is for use as a diluent in microbial limits testing.
Have Questions?
Catalog NumberFormatMedia TypeIncludes
R0760116mm x 125mm TubePhosphate Buffer-Butterfield's0.1% Peptone
Catalog number R07601
Price (USD)
-
Request A Quote
Format:
16mm x 125mm Tube
Media Type:
Phosphate Buffer-Butterfield's
Includes:
0.1% Peptone

Thermo Scientific™ Remel Butterfield's with 0.1% Peptone Phosphate Buffer is for use as a diluent in microbial limits testing.

Specifications
Format16mm x 125mm Tube
Includes0.1% Peptone
Media TypePhosphate Buffer-Butterfield's
Quantity100 x 9 mL
Product TypeBuffer Solution
Unit SizeEach