TopVision Low Melting Point Agarose
Thermo Scientific™

TopVision Low Melting Point Agarose

Perform in-gel enzymatic process experiments and nucleic acid recovery after electrophoresis with Thermo Scientific™ TopVision™ Low Melting Point Agarose.
Have Questions?
Catalog NumberQuantity
R080125 g
Catalog number R0801
Price (USD)
211.00
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
211.00
Each
Top Vision Low Melting Point Agarose is highly purified agarose with a low melting temperature certified by strict quality control test procedures. It is used for in-gel enzymatic process experiments and for nucleic acids recovery from gels using Agarase (#EO0461).

Thermo Scientific TopVision Low Melting Point Agarose is used for in-gel enzymatic process experiments and for nucleic acid recovery after electrophoresis.

Highlights

  • Optimal concentration between 0.7-2% in all typical buffer systems
  • GQ (Genetic Quality) certified, which ensures that nucleic acids recovered from preparative gels can be used for downstream applications (enzymatic reactions etc.)
  • Low DNA/RNA binding
  • Excellent gel transparency
  • DNase and RNase free
  • Suitable for RNA analysis

Applications

  • Analytical electrophoresis of nucleic acids
  • Preparative electrophoresis
  • In-gel enzymatic processing experiments
  • Preparation of DNA preparative gels in routine molecular biology techniques
  • The DNA recovered from agarose gels after electrophoresis can be used in enzymatic process (restriction, ligation, etc.)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Melting Point65.5°C
Quantity25 g
Validated ApplicationGel Electrophoresis
Product LineTopVision
Product TypeAgarose Gel
Separation Range100 to >30000 bp
Unit SizeEach