Thermo Scientific™

Inhibitory Mold Agar

Catalog number: R08412
Thermo Scientific™

Inhibitory Mold Agar

Catalog number: R08412

Thermo Scientific™ Remel™ Inhibitory Mold Agar (w/chloramphenicol) is used to primarily isolate and cultivate Histoplasma capsulatum, dermatophytes, and other pathogenic fungi. Ulrich formulated Inhibitory Mold Agar as a general-purpose medium for cultivation of fungi1. It is also recommended for selective isolation of pathogenic yeasts and molds such as Cryptococcus neoformans, H. capsulatum, and Trichophyton spp. from clinical specimens contaminated with commensal microbial flora2,3.

 
Catalog Number
R08412
Unit Size
Each
Format
20mm x 113mm Tube
Quantity
20/Pk.
Price (USD)
Full specifications
DescriptionInhibitory Mold Agar
Format20mm x 113mm Tube
Product TypeAgar
TypeAgar
Quantity20/Pk.
Unit SizeEach
Catalog Number
Unit Size
Each
Format
Monoplate - 26mL
Quantity
10 x 26 mL
Price (USD)
Full specifications
DescriptionInhibitory Mold Agar
FormatMonoplate - 26mL
Product TypeAgar
TypeAgar
Quantity10 x 26 mL
Unit SizeEach
Catalog Number
Unit Size
Each
Format
Monoplate - 35mL
Quantity
10 x 35 mL
Price (USD)
Full specifications
DescriptionInhibitory Mold Agar
FormatMonoplate - 35mL
Product TypeAgar
TypeAgar
Quantity10 x 35 mL
Unit SizeEach
Catalog Number
Unit Size
Each
Format
20mm x 113mm Tube
Quantity
100/Pk.
Price (USD)
Full specifications
DescriptionInhibitory Mold Agar
Format20mm x 113mm Tube
Product TypeAgar
TypeAgar
Quantity100/Pk.
Unit SizeEach
Catalog Number
Unit Size
Each
Format
1 oz. Bottle
Quantity
100/Pk.
Price (USD)
Full specifications
DescriptionInhibitory Mold Agar
Format1 oz. Bottle
Product TypeAgar
TypeAgar
Quantity100/Pk.
Unit SizeEach
Showing 5 of 5
Catalog NumberSpecificationsUnit SizeFormatQuantityPrice (USD)
R08412Full specifications
Each20mm x 113mm Tube20/Pk.Request A Quote
DescriptionInhibitory Mold Agar
Format20mm x 113mm Tube
Product TypeAgar
TypeAgar
Quantity20/Pk.
Unit SizeEach
R01505Full specifications
EachMonoplate - 26mL10 x 26 mLRequest A Quote
DescriptionInhibitory Mold Agar
FormatMonoplate - 26mL
Product TypeAgar
TypeAgar
Quantity10 x 26 mL
Unit SizeEach
R01508Full specifications
EachMonoplate - 35mL10 x 35 mLRequest A Quote
DescriptionInhibitory Mold Agar
FormatMonoplate - 35mL
Product TypeAgar
TypeAgar
Quantity10 x 35 mL
Unit SizeEach
R08410Full specifications
Each20mm x 113mm Tube100/Pk.Request A Quote
DescriptionInhibitory Mold Agar
Format20mm x 113mm Tube
Product TypeAgar
TypeAgar
Quantity100/Pk.
Unit SizeEach
R10070Full specifications
Each1 oz. Bottle100/Pk.Request A Quote
DescriptionInhibitory Mold Agar
Format1 oz. Bottle
Product TypeAgar
TypeAgar
Quantity100/Pk.
Unit SizeEach
Showing 5 of 5

Inhibitory Mold Agar primarily supports isolation and cultivation of dermatophytes and other pathogenic fungi. Inhibitory Mold Agar contains casein and meat peptones that supply nitrogenous compounds, amino acids, and peptides necessary for the growth of fungi. Dextrose, starch, and dextrin are ready sources of energy. Sodium chloride and metallic salts are essential electrolytes that help to maintains osmotic equilibrium. Yeast extract is a source of vitamins.

  • Ready to use - Solid media
  • Recommended for selective isolation of pathogenic yeasts and molds such as Cryptococcus neoformans, capsulatum, and Trichophyton spp. from clinical specimens contaminated with commensal microbial flora2,3
  • Easy to read - Examine plates for fungal colonies that exhibit typical color and morphology
  • Selective media - Chloramphenicol inhibits many gram-positive and gram-negative bacteria while gentamicin, ciprofloxacin, and vancomycin inhibit commensal microbial flora

Figures