Thermo Scientific™
Remel™ Lowenstein-Jensen Medium w/5% NaCl (LJ)
 Related applications:
Clinical Microbiology
Thermo Scientific™ Remel Lowenstein-Jensen Medium with 5% NaCl (LJ) is for the detection of sodium chloride tolerance to aid in the differentiation of Mycobacterium spp.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
Thermo Scientific™ Remel Lowenstein-Jensen Medium with 5% NaCl (LJ) is for the detection of sodium chloride tolerance to aid in the differentiation of Mycobacterium spp.
Documents
Manuals & protocols