X-Gal Solution, ready-to-use
Thermo Scientific™
X-Gal Solution, ready-to-use
Thermo Scientific X-Gal (5-bromo-4-chloro-3-indolyl-beta-D-galacto-pyranoside) is an inert chromogenic substrate for beta-galactosidase which hydrolyzes X-Gal into colorless galactose and 4-chloro-3-brom-indigo, formingRead more
Have Questions?
Catalog NumberQuantity
R094110 mL
Catalog number R0941
Price (KRW)
161,000
Online offer
Ends: 30-Jun-2026
183,000
Save 22,000 (12%)
Each
Quantity:
10 mL
Request bulk or custom format
Price (KRW)
161,000
Online offer
Ends: 30-Jun-2026
183,000
Save 22,000 (12%)
Each
Thermo Scientific X-Gal (5-bromo-4-chloro-3-indolyl-beta-D-galacto-pyranoside) is an inert chromogenic substrate for beta-galactosidase which hydrolyzes X-Gal into colorless galactose and 4-chloro-3-brom-indigo, forming an intense blue precipitate. Induction of the lacZ gene with IPTG leads to the hydrolysis of X-Gal and to the development of blue colonies (see Supporting data).
X-Gal Solution, ready-to-use, is stable, 0.22 μm membrane filtered solution formulated for direct use in conjunction with IPTG for blue/white colony screening.
Applications
• Blue/white colony screening to distinguish recombinant (white) from non-recombinant (blue) colonies
• Visualization of beta-galactosidase reporter gene expression in transfected eukaryotic cells
• Detection of beta-galactosidase activity in immunological and histochemical procedures
Related Products
X-Gal
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypeX-Gal Solution
Quantity10 mL
Substrate PropertiesChemical Substrate
Substrate TypeBeta-Gal Substrate, X-Gal
Detection MethodColorimetric
Unit SizeEach