Thermo Scientific™

Plate Count Agar

Catalog number: R09640
Thermo Scientific™

Plate Count Agar

Catalog number: R09640
Enumerate viable organisms in milk, dairy products, water, food and other specimens of sanitary importance using non-selective Thermo Scientific™ Plate Count Agar.
 
Catalog Number
R09640
Unit Size
Each
Description
Plate Count Agar
Format
20mm x 150mm Tube
Quantity
100/Pk.
Manufacturing Origin
US
Price (USD)
Full specifications
Format20mm x 150mm Tube
TypeAgar
DescriptionPlate Count Agar
Product TypeAgar
Manufacturing OriginUS
Quantity100/Pk.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Plate Count Agar
Format
85mm Monoplate
Quantity
10/Pk.
Manufacturing Origin
US
Price (USD)
Full specifications
Format85mm Monoplate
TypeAgar
DescriptionPlate Count Agar
Product TypeAgar
Manufacturing OriginUS
Quantity10/Pk.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Plate Count Agar
Format
Tube - Slant
Quantity
20/Pk.
Manufacturing Origin
US
Price (USD)
Full specifications
FormatTube - Slant
TypeAgar
DescriptionPlate Count Agar
Product TypeAgar
Manufacturing OriginUS
Quantity20/Pk.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Plate Count Agar
Format
20mm x 150mm Tube
Quantity
20/Pk.
Manufacturing Origin
US
Price (USD)
Full specifications
Format20mm x 150mm Tube
TypeAgar
DescriptionPlate Count Agar
Product TypeAgar
Manufacturing OriginUS
Quantity20/Pk.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Plate Count Agar
Format
Bottle - 200mL
Quantity
10/Pk.
Manufacturing Origin
US
Price (USD)
Full specifications
FormatBottle - 200mL
TypeAgar
DescriptionPlate Count Agar
Product TypeAgar
Manufacturing OriginUS
Quantity10/Pk.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Plate Count Agar
Format
Bottle - 500mL
Quantity
10/Pk.
Manufacturing Origin
US
Price (USD)
Full specifications
FormatBottle - 500mL
TypeAgar
DescriptionPlate Count Agar
Product TypeAgar
Manufacturing OriginUS
Quantity10/Pk.
Unit SizeEach
Showing 6 of 6
Catalog NumberSpecificationsUnit SizeDescriptionFormatQuantityManufacturing OriginPrice (USD)
R09640Full specifications
EachPlate Count Agar20mm x 150mm Tube100/Pk.USRequest A Quote
Format20mm x 150mm Tube
TypeAgar
DescriptionPlate Count Agar
Product TypeAgar
Manufacturing OriginUS
Quantity100/Pk.
Unit SizeEach
R01685Full specifications
EachPlate Count Agar85mm Monoplate10/Pk.USRequest A Quote
Format85mm Monoplate
TypeAgar
DescriptionPlate Count Agar
Product TypeAgar
Manufacturing OriginUS
Quantity10/Pk.
Unit SizeEach
R062602Full specifications
EachPlate Count AgarTube - Slant20/Pk.USRequest A Quote
FormatTube - Slant
TypeAgar
DescriptionPlate Count Agar
Product TypeAgar
Manufacturing OriginUS
Quantity20/Pk.
Unit SizeEach
R09642Full specifications
EachPlate Count Agar20mm x 150mm Tube20/Pk.USRequest A Quote
Format20mm x 150mm Tube
TypeAgar
DescriptionPlate Count Agar
Product TypeAgar
Manufacturing OriginUS
Quantity20/Pk.
Unit SizeEach
R112590Full specifications
EachPlate Count AgarBottle - 200mL10/Pk.USRequest A Quote
FormatBottle - 200mL
TypeAgar
DescriptionPlate Count Agar
Product TypeAgar
Manufacturing OriginUS
Quantity10/Pk.
Unit SizeEach
R112591Full specifications
EachPlate Count AgarBottle - 500mL10/Pk.USRequest A Quote
FormatBottle - 500mL
TypeAgar
DescriptionPlate Count Agar
Product TypeAgar
Manufacturing OriginUS
Quantity10/Pk.
Unit SizeEach
Showing 6 of 6
This standard medium meets the formulation of APHA, AOAC and PHLS.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

Figures

Frequently asked questions (FAQs)

Citations & References