Search Thermo Fisher Scientific
Thermo Scientific™ Remel TOC Agar (Polysorbate 80, Oxgall, Caffeic Acid) is for the presumptive identification of Candida albicans and Cryptococcus neoformans by germ tube formation, chlamydospore production and phenol oxidase activity.
Description | TOC Agar |
Format | 20mm x 150mm Pour Tube |
Product Type | Agar |
Type | Agar |
Quantity | 20/Pk. |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Price (USD) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
R09892 | Full specifications | Each | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Thermo Scientific™ Remel TOC Agar (Polysorbate 80, Oxgall, Caffeic Acid) is for the presumptive identification of Candida albicans and Cryptococcus neoformans by germ tube formation, chlamydospore production and phenol oxidase activity.