Thermo Scientific™ Remel Trichophyton Agar provides differentiation of Trichophyton spp. based on nutritional requirements.
Format | G - PT |
Type | Agar |
Description | Trichophyton Agar No. 1 |
Product Type | Agar |
Quantity | 20/Pk. |
Unit Size | Each |
Format | G - PT |
Type | Agar |
Description | Trichophyton Agar No. 7 |
Product Type | Agar |
Quantity | 20/Pk. |
Unit Size | Each |
Format | G - PT |
Type | Agar |
Description | Trichophyton Agar No. 6 |
Product Type | Agar |
Quantity | 20/Pk. |
Unit Size | Each |
Description | Trichophyton Agar No. 5 |
Format | G - PT |
Product Type | Agar |
Type | Agar |
Quantity | 20/Pk. |
Unit Size | Each |
Description | Trichophyton Agar No. 4 |
Format | 20mm x 150mm Pour Tube |
Product Type | Agar |
Type | Agar |
Quantity | 20/Pk. |
Unit Size | Each |
Description | Trichophyton Agar No. 3 |
Format | 20mm x 150mm Pour Tube |
Product Type | Agar |
Type | Agar |
Quantity | 20/Pk. |
Unit Size | Each |
Description | Trichophyton Agar No. 2 |
Format | 20mm x 150mm Pour Tube |
Product Type | Agar |
Type | Agar |
Quantity | 20/Pk. |
Unit Size | Each |
Description | Trichophyton Agar No. 6 |
Format | 20mm x 113mm Tube - Slant |
Product Type | Agar |
Type | Agar |
Quantity | 20/Pk. |
Unit Size | Each |
Description | Trichophyton Agar No. 5 |
Format | 20mm x 113mm Tube - Slant |
Product Type | Agar |
Type | Agar |
Quantity | 20/Pk. |
Unit Size | Each |
Format | 20mm x 113mm Tube - Slant |
Type | Agar |
Description | Trichophyton Agar No. 4 |
Product Type | Agar |
Quantity | 20/Pk. |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Format | Price (USD) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
R09903 | Each | Trichophyton Agar No. 1 | G - PT | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R09927 | Each | Trichophyton Agar No. 7 | G - PT | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R09923 | Each | Trichophyton Agar No. 6 | G - PT | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R09919 | Each | Trichophyton Agar No. 5 | G - PT | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R09915 | Each | Trichophyton Agar No. 4 | 20mm x 150mm Pour Tube | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R09911 | Each | Trichophyton Agar No. 3 | 20mm x 150mm Pour Tube | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R09907 | Each | Trichophyton Agar No. 2 | 20mm x 150mm Pour Tube | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R08921 | Each | Trichophyton Agar No. 6 | 20mm x 113mm Tube - Slant | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R08917 | Each | Trichophyton Agar No. 5 | 20mm x 113mm Tube - Slant | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R08913 | Each | Trichophyton Agar No. 4 | 20mm x 113mm Tube - Slant | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Thermo Scientific™ Remel Trichophyton Agar provides differentiation of Trichophyton spp. based on nutritional requirements.