Thermo Scientific™

Brain Heart Infusion (BHI) Agar

Catalog number: R10034
Thermo Scientific™

Brain Heart Infusion (BHI) Agar

Catalog number: R10034

Isolate and cultivate a wide variety of microorganisms including pathogenic fungi with Thermo Scientific™ Remel™ Brain Heart Infusion (BHI) Agar.  Due to the addition of 5-10% sheep blood, BHI agar can be used for the isolation of dimorphic fungi which may grow poorly on non-enriched media. BHI Agar is recommended by the Association of Official Analytical Chemists (AOAC) and the American Public Health Association (APHA) 1, 2

 
Catalog Number
R10034
Unit Size
Each
Format
1 oz. Bottle
Quantity
100/Pk.
Price (USD)
Full specifications
Format1 oz. Bottle
TypeAgar
DescriptionBrain Heart Infusion Agar (BHI)
Product TypeAgar
Manufacturing OriginUS
Quantity100/Pk.
Certifications/ComplianceIndustrial Reference: BAM, COMPF, EPA, IDF, SMD.
Unit SizeEach
Catalog Number
Unit Size
Each
Format
15mm x 103mm Tube
Quantity
20/Pk.
Price (USD)
Full specifications
Format15mm x 103mm Tube
TypeAgar
DescriptionBHI Agar (Brain Heart Infusion Agar)
Product TypeAgar
Quantity20/Pk.
Unit SizeEach
Catalog Number
Unit Size
Each
Format
20mm x 113mm Tube - Slant
Quantity
100/Pk.
Price (USD)
Full specifications
Format20mm x 113mm Tube - Slant
TypeAgar
DescriptionBrain Heart Infusion Agar (BHI)
Product TypeAgar
Manufacturing OriginUS
Quantity100/Pk.
Certifications/ComplianceIndustrial Reference: BAM, COMPF, EPA, IDF, SMD.
Unit SizeEach
Catalog Number
Unit Size
Each
Format
20mm x 113mm Tube - Slant
Quantity
20/Pk.
Price (USD)
Full specifications
Format20mm x 113mm Tube - Slant
TypeAgar
DescriptionBrain Heart Infusion Agar (BHI)
Product TypeAgar
Manufacturing OriginUS
Quantity20/Pk.
Certifications/ComplianceIndustrial Reference: BAM, COMPF, EPA, IDF, SMD.
Unit SizeEach
Catalog Number
Unit Size
Each
Format
20mm x 150mm Pour Tube
Quantity
20/Pk.
Price (USD)
Full specifications
Format20mm x 150mm Pour Tube
TypeAgar
DescriptionBrain Heart Infusion Agar (BHI)
Product TypeAgar
Manufacturing OriginUS
Quantity20/Pk.
Certifications/ComplianceIndustrial Reference: BAM, COMPF, EPA, IDF, SMD.
Unit SizeEach
Showing 5 of 5
Catalog NumberSpecificationsUnit SizeFormatQuantityPrice (USD)
R10034Full specifications
Each1 oz. Bottle100/Pk.Request A Quote
Format1 oz. Bottle
TypeAgar
DescriptionBrain Heart Infusion Agar (BHI)
Product TypeAgar
Manufacturing OriginUS
Quantity100/Pk.
Certifications/ComplianceIndustrial Reference: BAM, COMPF, EPA, IDF, SMD.
Unit SizeEach
R060232Full specifications
Each15mm x 103mm Tube20/Pk.Request A Quote
Format15mm x 103mm Tube
TypeAgar
DescriptionBHI Agar (Brain Heart Infusion Agar)
Product TypeAgar
Quantity20/Pk.
Unit SizeEach
R08060Full specifications
Each20mm x 113mm Tube - Slant100/Pk.Request A Quote
Format20mm x 113mm Tube - Slant
TypeAgar
DescriptionBrain Heart Infusion Agar (BHI)
Product TypeAgar
Manufacturing OriginUS
Quantity100/Pk.
Certifications/ComplianceIndustrial Reference: BAM, COMPF, EPA, IDF, SMD.
Unit SizeEach
R08062Full specifications
Each20mm x 113mm Tube - Slant20/Pk.Request A Quote
Format20mm x 113mm Tube - Slant
TypeAgar
DescriptionBrain Heart Infusion Agar (BHI)
Product TypeAgar
Manufacturing OriginUS
Quantity20/Pk.
Certifications/ComplianceIndustrial Reference: BAM, COMPF, EPA, IDF, SMD.
Unit SizeEach
R09060Full specifications
Each20mm x 150mm Pour Tube20/Pk.Request A Quote
Format20mm x 150mm Pour Tube
TypeAgar
DescriptionBrain Heart Infusion Agar (BHI)
Product TypeAgar
Manufacturing OriginUS
Quantity20/Pk.
Certifications/ComplianceIndustrial Reference: BAM, COMPF, EPA, IDF, SMD.
Unit SizeEach
Showing 5 of 5

Use Heart Infusion Agar for the cultivation of a wide variety of microorganisms including pathogenic fungi.

  • Recommended by AOAC and APHA.
  • Ready-to-use convenience of prepared media.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

REF R112025 not intended for IVD use.
For In Vitro Diagnostic Use.  

Figures

Frequently asked questions (FAQs)

Citations & References