Thermo Scientific™

Sabouraud Dextrose Agar

Catalog number: R10156
Thermo Scientific™

Sabouraud Dextrose Agar

Catalog number: R10156
Isolate, cultivate and differentiate dermatophytes, fungi and yeasts, such as Candida albicans, using acidic pH Thermo Scientific™ Sabouraud Glucose Agar.
 
Catalog Number
R10156
Unit Size
Each
Description
Sabouraud Dextrose Agar, pH 5.6
Format
1 oz. Bottle
Quantity
10/Pk.
Manufacturing Origin
US
Price (USD)
Full specifications
Fill Volume1 oz.
Shelf Life36-52 Weeks
Format1 oz. Bottle
TypeAgar
ColorAmber
DescriptionSabouraud Dextrose Agar, pH 5.6
Product TypeAgar
Manufacturing OriginUS
Quantity10/Pk.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Sabouraud Dextrose Agar, pH 5.6
Format
Monoplate Deep Pour
Quantity
100/Pk.
Manufacturing Origin
US
Price (USD)
Full specifications
Fill Volume26 mL
Shelf Life8-12 Weeks
FormatMonoplate Deep Pour
TypeAgar
ColorAmber
DescriptionSabouraud Dextrose Agar, pH 5.6
Product TypeAgar
Manufacturing OriginUS
Quantity100/Pk.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Sabouraud Dextrose Agar, pH 5.6
Format
Monoplate Deep Pour
Quantity
10/Pk.
Manufacturing Origin
US
Price (USD)
Full specifications
Fill Volume26 mL
Shelf Life8-12 Weeks
FormatMonoplate Deep Pour
TypeAgar
ColorAmber
DescriptionSabouraud Dextrose Agar, pH 5.6
Product TypeAgar
Manufacturing OriginUS
Quantity10/Pk.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Sabouraud Dextrose Agar, pH 5.6
Format
20mm x 150mm Tube - Slant
Quantity
100/Pk.
Manufacturing Origin
US
Price (USD)
Full specifications
Fill Volume10 mL
Shelf Life8-12 Weeks
Format20mm x 150mm Tube - Slant
TypeAgar
ColorAmber
DescriptionSabouraud Dextrose Agar, pH 5.6
Product TypeAgar
Manufacturing OriginUS
Quantity100/Pk.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Sabouraud Dextrose Agar, pH 5.6
Format
20mm x 150mm Tube - Slant
Quantity
20/Pk.
Manufacturing Origin
US
Price (USD)
Full specifications
Fill Volume10 mL
Shelf Life8-12 Weeks
Format20mm x 150mm Tube - Slant
TypeAgar
ColorAmber
DescriptionSabouraud Dextrose Agar, pH 5.6
Product TypeAgar
Manufacturing OriginUS
Quantity20/Pk.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Sabouraud Dextrose Agar, pH 5.6
Format
1 oz. Bottle
Quantity
100/Pk.
Manufacturing Origin
US
Price (USD)
Full specifications
Fill Volume1 oz.
Shelf Life36-52 Weeks
Format1 oz. Bottle
TypeAgar
ColorAmber
DescriptionSabouraud Dextrose Agar, pH 5.6
Product TypeAgar
Manufacturing OriginUS
Quantity100/Pk.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Sabouraud Dextrose Agar, pH 5.6
Format
BB - 200mL
Quantity
10/Pk.
Manufacturing Origin
US
Price (USD)
Full specifications
Fill Volume200 mL
Shelf Life36-52 Weeks
FormatBB - 200mL
TypeAgar
ColorAmber
DescriptionSabouraud Dextrose Agar, pH 5.6
Product TypeAgar
Manufacturing OriginUS
Quantity10/Pk.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Sabouraud Dextrose Agar, pH 5.6
Format
BB - 500mL
Quantity
10/Pk.
Manufacturing Origin
US
Price (USD)
Full specifications
Fill Volume500 mL
Shelf Life36-52 Weeks
FormatBB - 500mL
TypeAgar
ColorAmber
DescriptionSabouraud Dextrose Agar, pH 5.6
Product TypeAgar
Manufacturing OriginUS
Quantity10/Pk.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Sabouraud Dextrose Agar, pH 5.6
Format
BRB - 100mL
Quantity
10/Pk.
Manufacturing Origin
US
Price (USD)
Full specifications
Fill Volume100 mL
Packaging Type10 x 100mL
Shelf Life36-52 Weeks
FormatBRB - 100mL
TypeAgar
ColorAmber
DescriptionSabouraud Dextrose Agar, pH 5.6
Product TypeAgar
Manufacturing OriginUS
Quantity10/Pk.
Certifications/ComplianceIndustrial Reference: BAM, COMPF, USP
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Sabouraud Dextrose Agar, pH 5.6
Format
20 x 150mm Pour Tube (G-PT)
Quantity
100/Pk.
Manufacturing Origin
US
Price (USD)
Full specifications
Fill Volume20 mL
Shelf Life36-52 Weeks
Format20 x 150mm Pour Tube (G-PT)
TypeAgar
ColorAmber
DescriptionSabouraud Dextrose Agar, pH 5.6
Product TypeAgar
Manufacturing OriginUS
Quantity100/Pk.
Certifications/ComplianceIndustrial Reference: BAM, COMPF, USP.
Unit SizeEach
Showing 10 of 10
Catalog NumberSpecificationsUnit SizeDescriptionFormatQuantityManufacturing OriginPrice (USD)
R10156Full specifications
EachSabouraud Dextrose Agar, pH 5.61 oz. Bottle10/Pk.USRequest A Quote
Fill Volume1 oz.
Shelf Life36-52 Weeks
Format1 oz. Bottle
TypeAgar
ColorAmber
DescriptionSabouraud Dextrose Agar, pH 5.6
Product TypeAgar
Manufacturing OriginUS
Quantity10/Pk.
Unit SizeEach
R01766Full specifications
EachSabouraud Dextrose Agar, pH 5.6Monoplate Deep Pour100/Pk.USRequest A Quote
Fill Volume26 mL
Shelf Life8-12 Weeks
FormatMonoplate Deep Pour
TypeAgar
ColorAmber
DescriptionSabouraud Dextrose Agar, pH 5.6
Product TypeAgar
Manufacturing OriginUS
Quantity100/Pk.
Unit SizeEach
R01768Full specifications
EachSabouraud Dextrose Agar, pH 5.6Monoplate Deep Pour10/Pk.USRequest A Quote
Fill Volume26 mL
Shelf Life8-12 Weeks
FormatMonoplate Deep Pour
TypeAgar
ColorAmber
DescriptionSabouraud Dextrose Agar, pH 5.6
Product TypeAgar
Manufacturing OriginUS
Quantity10/Pk.
Unit SizeEach
R09746Full specifications
EachSabouraud Dextrose Agar, pH 5.620mm x 150mm Tube - Slant100/Pk.USRequest A Quote
Fill Volume10 mL
Shelf Life8-12 Weeks
Format20mm x 150mm Tube - Slant
TypeAgar
ColorAmber
DescriptionSabouraud Dextrose Agar, pH 5.6
Product TypeAgar
Manufacturing OriginUS
Quantity100/Pk.
Unit SizeEach
R09747Full specifications
EachSabouraud Dextrose Agar, pH 5.620mm x 150mm Tube - Slant20/Pk.USRequest A Quote
Fill Volume10 mL
Shelf Life8-12 Weeks
Format20mm x 150mm Tube - Slant
TypeAgar
ColorAmber
DescriptionSabouraud Dextrose Agar, pH 5.6
Product TypeAgar
Manufacturing OriginUS
Quantity20/Pk.
Unit SizeEach
R10155Full specifications
EachSabouraud Dextrose Agar, pH 5.61 oz. Bottle100/Pk.USRequest A Quote
Fill Volume1 oz.
Shelf Life36-52 Weeks
Format1 oz. Bottle
TypeAgar
ColorAmber
DescriptionSabouraud Dextrose Agar, pH 5.6
Product TypeAgar
Manufacturing OriginUS
Quantity100/Pk.
Unit SizeEach
R112550Full specifications
EachSabouraud Dextrose Agar, pH 5.6BB - 200mL10/Pk.USRequest A Quote
Fill Volume200 mL
Shelf Life36-52 Weeks
FormatBB - 200mL
TypeAgar
ColorAmber
DescriptionSabouraud Dextrose Agar, pH 5.6
Product TypeAgar
Manufacturing OriginUS
Quantity10/Pk.
Unit SizeEach
R112551Full specifications
EachSabouraud Dextrose Agar, pH 5.6BB - 500mL10/Pk.USRequest A Quote
Fill Volume500 mL
Shelf Life36-52 Weeks
FormatBB - 500mL
TypeAgar
ColorAmber
DescriptionSabouraud Dextrose Agar, pH 5.6
Product TypeAgar
Manufacturing OriginUS
Quantity10/Pk.
Unit SizeEach
R112562Full specifications
EachSabouraud Dextrose Agar, pH 5.6BRB - 100mL10/Pk.USRequest A Quote
Fill Volume100 mL
Packaging Type10 x 100mL
Shelf Life36-52 Weeks
FormatBRB - 100mL
TypeAgar
ColorAmber
DescriptionSabouraud Dextrose Agar, pH 5.6
Product TypeAgar
Manufacturing OriginUS
Quantity10/Pk.
Certifications/ComplianceIndustrial Reference: BAM, COMPF, USP
Unit SizeEach
R09744Full specifications
EachSabouraud Dextrose Agar, pH 5.620 x 150mm Pour Tube (G-PT)100/Pk.USRequest A Quote
Fill Volume20 mL
Shelf Life36-52 Weeks
Format20 x 150mm Pour Tube (G-PT)
TypeAgar
ColorAmber
DescriptionSabouraud Dextrose Agar, pH 5.6
Product TypeAgar
Manufacturing OriginUS
Quantity100/Pk.
Certifications/ComplianceIndustrial Reference: BAM, COMPF, USP.
Unit SizeEach
Showing 10 of 10
Fungi maintain their typical cultural appearance and may thus be readily identified according to the standard macroscopic characteristics. Infections caused by Candida spp. are diverse, ranging from superficial and local to deep-seated and disseminated. Candida albicans and Candida glabrata comprise 70-80% of yeast isolates recovered from patients with invasive disease. This culture media, also available in powder form, has applications in environmental monitoring and sterility testing, enabling corrective measures to be implemented rapidly if unacceptable levels of microbial loading indicate lapses in process control.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

Contents & Storage

2°C to 25°C

Figures

Frequently asked questions (FAQs)

Citations & References