Image RNA at single-molecule sensitivity and single-cell resolution with ViewRNA Cell Plus Assay.
Invitrogen™
Molecular Probes® Cell Imaging Kit
Catalog number:  R10477
 Related applications:
Cellular Imaging
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
Fluorescence imaging protocols are sometimes complicated and tedious, with more time spent preparing samples and reagents than acquiring images and data. To help researchers focus more on biology and get results faster, we have developed a set of imaging reagents that emphasize user friendliness and workflow convenience.
The Molecular Probes® Cell Imaging Kit contains the following products:
NucBlue™ Live ReadyProbes™ Reagent (Cat. No. R37605)
NucRed™ Live 647 ReadyProbes™ Reagent (Cat. No. R37106)
ReadyProbes™ Cell Viability Imaging Kit, Blue/Green (Cat. No. R37609)
Goat Anti-Rabbit IgG (H+L) Secondary Antibody, Alexa Fluor™ 594 conjugate (Cat. No. R37117)
Goat Anti-Mouse lgG Secondary Antibody, Alexa Fluor™ 488 conjugate (Cat. No. R37120)
Image-iT™ FX Signal Enhancer ReadyProbes™ Reagent (Cat. No. R37107)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Reagent Type: Anti-Fade Solution, Microscopy Contrast Agent, Mounting Solution, Signal Enhancers
Product Line: ActinGreen™, BackDrop™, Image-iT®, NucBlue®, ProLong®, ReadyProbes®, SlowFade®
Product Size: 1 kit
Contents & storage
Store at room temperature.
Protect from light.
Do not freeze.
Documents
Manuals & protocols