Aminoallyl-dUTP Solution (50 mM)
Thermo Scientific™

Aminoallyl-dUTP Solution (50 mM)

Thermo Scientific Aminoallyl-dUTP (5-[3-aminoallyl]-2'-deoxyuridine-5'-triphosphate) is supplied as 10 mM and 50 mM aqueous solutions titrated to pH 7.0 with NaOH.Read more
Have Questions?
Catalog NumberQuantity
R110150 μL
Catalog number R1101
Price (USD)
199.65
Online exclusive
243.00
Save 43.35
Each
Quantity:
50 μL
Request bulk or custom format
Price (USD)
199.65
Online exclusive
243.00
Save 43.35
Each

Thermo Scientific Aminoallyl-dUTP (5-[3-aminoallyl]-2'-deoxyuridine-5'-triphosphate) is supplied as 10 mM and 50 mM aqueous solutions titrated to pH 7.0 with NaOH. Aminoallyl-dUTP can be enzymatically incorporated into DNA and the resulting amine-containing DNA can be subsequently labeled with any amine-reactive fluorescent dye, biotin or hapten.

Applications
• Enzymatic indirect non-radioactive labeling of DNA during cDNA synthesis
• PCR
• Nick-translation
• Random-primed labeling
• Primer extension

Aminoallyl-dUTP can be enzymatically incorporated into DNA with:
• Reverse transcriptases
• Taq DNA polymerase
• phi29 DNA Polymerase
• Klenow Fragment
• Klenow Fragment, exo-
• DNA Polymerase I

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Labeling MethodDirect Labeling
PolymeraseDNA Polymerase
Quantity50 μL
Shipping ConditionDry Ice
Concentration50 mM
For Use With (Application)Standard PCR, Gene Expression, Non-Radioactive Labeling
FormLiquid
Label or DyeAminoallyl
Unit SizeEach
Contents & Storage
Aminoallyl-dUTP Solution, 50 mM (50 μL)

Store at –20°C.