Product Image
Thermo Scientific™
Remel™ Contact I Rose Bengal Agar
Thermo Scientific™ Remel Contact I Rose Bengal Agar is used for selective isolation and enumeration of yeast, and fungi from environmental samples and food.
Have Questions?
Catalog NumberdimFormat
R111042
Catalog number R111042
Price (KRW)
-
Thermo Scientific™ Remel Contact I Rose Bengal Agar is used for selective isolation and enumeration of yeast, and fungi from environmental samples and food.
Specifications
DescriptionContact I Rose Bengal Agar w/chloramphenicol
FormatContact - Single Bag
Quantity20/Pk.
Product TypeAgar Plate
Unit SizeEach