Thermo Scientific™
Remel™ Contact I D/E (Dey-Engley) Neutralizing Agar
Catalog number: R111103
Thermo Scientific™
Remel™ Contact I D/E (Dey-Engley) Neutralizing Agar
Catalog number: R111103
Thermo Scientific™ Remel Contact I D/E Neutralizing Agar is used for the cultivation of microorganisms with neutralization of specific chemical disinfectants.
Catalog Number
R111103
Unit Size
Each
Quantity
20/Pk.
Price (USD)
Full specifications
FormatContact - Single Bag
TypeAgar
DescriptionContact I D/E Neutralizing Agar
Product TypeAgar
Quantity20/Pk.
Unit SizeEach
Catalog Number
Unit Size
Each
Quantity
100/Pk.
Price (USD)
Full specifications
FormatContact - Single Bag
TypeAgar
DescriptionContact I D/E Neutralizing Agar
Product TypeAgar
Quantity100/Pk.
Unit SizeEach
Showing 2 of 2
Catalog NumberSpecificationsUnit SizeQuantityPrice (USD)
R111103Full specifications
Each20/Pk.Request A Quote
FormatContact - Single Bag
TypeAgar
DescriptionContact I D/E Neutralizing Agar
Product TypeAgar
Quantity20/Pk.
Unit SizeEach
R111104Full specifications
Each100/Pk.Request A Quote
FormatContact - Single Bag
TypeAgar
DescriptionContact I D/E Neutralizing Agar
Product TypeAgar
Quantity100/Pk.
Unit SizeEach
Showing 2 of 2
  • Manufactured in a clean room environment to provide the highest level of sterility assurance
  • Made using GripFit™ technology, which ensures the lid remains in place during handling and yet can be easily removed during environmental monitoring
  • Contact I (Single Bag) and Contact II (Double Bag) are manufactured in a clean room environment
  • This provides the highest level of sterility assurance with no compromise of growth characteristics
  • Each lot is tested to ensure consistent, reproducible results
Not intended for IVD use.