Remel™ Contact Plate Sterile Tryptic Soy Agar w/Lecithin, Polysorbate 80 (Irradiated)
Remel™ Contact Plate Sterile Tryptic Soy Agar w/Lecithin, Polysorbate 80 (Irradiated)
Remel™ Contact Plate Sterile Tryptic Soy Agar w/Lecithin, Polysorbate 80 (Irradiated)
Thermo Scientific™
Remel™ Contact Plate Sterile Tryptic Soy Agar w/Lecithin, Polysorbate 80 (Irradiated)
Thermo Scientific™ Remel Contact Plate Sterile Tryptic Soy Agar with Lecithin, Polysorbate 80 (Irradiated) is for the cultivation of bacteria, yeast and fungi.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
R111802100/Pk.
R11180010/Pk.
Catalog number R111802
Price (USD)
-
Quantity:
100/Pk.
Thermo Scientific™ Remel Contact Plate Sterile Tryptic Soy Agar with Lecithin, Polysorbate 80 (Irradiated) is for the cultivation of bacteria, yeast and fungi.
  • Double-bagged plus a third bag is enclosed for customer use (Double + Bag)
  • Products are manufactured in a clean room and validated sterile by gamma radiation
  • Made using GripFit™ technology, which ensures the lid remains in place during handling and yet can be easily removed during environmental monitoring
Not intended for IVD use.
Specifications
DescriptionContact Plates; Sterile; Tryptic Soy Agar w/Lecithin; Gamma irradiated
FormatContact - Double Bag
Quantity100/Pk.
Product TypeAgar Plate
Unit SizeEach