Remel™ Isolator Wrap™ Sterile Sabauroud Dextrose Agar Contact Plate w/Lecithin, Polysorbate 80
Actual product may vary
Thermo Scientific™
Remel™ Isolator Wrap™ Sterile Sabauroud Dextrose Agar Contact Plate w/Lecithin, Polysorbate 80
Thermo Scientific™ Remel Isolator Wrap Sterile Sabauroud Dextrose Agar Contact Plate w/Lecithin, Polysorbate 80 is used for the cultivation of yeasts and fungi.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
R111826100/Pk.
R11182510/Pk.
Catalog number R111826
Price (USD)
-
Quantity:
100/Pk.
Thermo Scientific™ Remel Isolator Wrap Sterile Sabauroud Dextrose Agar Contact Plate w/Lecithin, Polysorbate 80 is used for the cultivation of yeasts and fungi.
  • Foil Barrier Wrap — impermeable to isolator decontamination process
  • GripFit™ Plates — easy to handle with one hand without lid separating from bottom of plate
  • Irradiated — Sterility Assurance Level 10-6
  • Shrink-wraped with easy open tear strip — reduces breakage and moisture loss
  • Bottom labeling — information stays with sample
  • Extra sterile bag — easy to transport plates to lab without introducing contamination
  • Six month shelf life — less ordering and fewer lot numbers to QC
Not intended for IVD use.
Specifications
FormatFoil Barrier Wrap + Bag
Quantity100/Pk.
Product TypeAgar Plate
Unit SizeEach