Thermo Scientific™
Remel™ Settling Plate Sterile Tryptic Soy Agar (Irradiated)
Related applications:
Industrial Microbiology
Thermo Scientific™ Remel Settling Plate Sterile Tryptic Soy Agar (Irradiated) is used for the cultivation of bacteria, yeast and fungi.
Back to top
  • Settling plates (single bag and double bag) are manufactured in a clean room environment
  • This provides the highest level of sterility assurance with no compromise of growth characteristics
  • Each lot is tested to ensure consistent, reproducible results
  • Plate size:84mm
Not intended for IVD use.