Triple Wrap Sterile Pack Tryptone Soya Agar with Neutralizers, VHP indicator, and 2D Barcode
Thermo Scientific™

Triple Wrap Sterile Pack Tryptone Soya Agar with Neutralizers, VHP indicator, and 2D Barcode

Carry out environmental monitoring in your most regulated environments using Triple Wrapped Plates, for passive and active air sampling and surface testing applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFormat
R111TW285 mm plate
R111TW155 mm plates
Catalog number R111TW2
Price (MXN)
-
Format:
85 mm plate

Experience next-level cleanroom confidence with Thermo Scientific™ Triple Wrap Sterile Pack Tryptone Soya Agar plates, complete with VHP indicator, Lecithin, Polysorbate 80, Sodium Thiosulphate and L-Histidin and 2D Barcode.

Achieve confidence in your results and ensure optimum media growth assurance using plates with a Vapour-phase Hydrogen Peroxide (VHP) exposure indicator, designed to confirm the integrity of the wrapping material and seal during VHP exposure.

Three protective layers reduce risk of contamination:

  • Primary film with moisture control patch
  • Secondary film VHP barrier with immobilized desiccant sachet
  • Tertiary film dust cover
  • Conveniently sold in packs of 100 which are sealed for security

The incorporation of novel elements enhance moisture control by permitting the passage and absorption of excess moisture

Using plates with an improved shelf life and room temperature storage offers a cost-effective solution, minimizing new batch validation and refrigeration costs

Not all products are available for sale in all territories. Please inquire.

Specifications
ColorIvory, transparent
DescriptionThermo Scientific™ Sterile Triple wrapped Tryptic Soy Agar settling with VHP indicator
Fill Weight30 grams (+/- 0.5)
Format85 mm plate
Packaging TypeNo cardboard inner packaging, triple wrapped-irradiated, with VHP barrier wrap
Quantity100 /pk
Shelf Life43 weeks
Product TypeAgar Plate
Unit SizeEach