Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Thermo Scientific™ Remel BHI Broth (Brain Heart Infusion Broth) is for the cultivation of microorganisms.
Certifications/Compliance | Industrial Reference: BAM, SMD, SMWW, USDA. |
Description | BHI Broth |
Format | Borosilicate Bottle |
Product Type | Microbiology Media |
Quantity | 10 x 500 mL |
Unit Size | Each |
Certifications/Compliance | Industrial Reference: BAM, SMD, SMWW, USDA. |
Description | BHI Broth |
Format | 15mm x 103mm Tube |
Product Type | Microbiology Media |
Quantity | 100 x 2 mL |
Unit Size | Each |
Certifications/Compliance | Industrial Reference: BAM, SMD, SMWW, USDA. |
Description | BHI Broth |
Format | 15mm x 103mm Tube |
Product Type | Microbiology Media |
Quantity | 100 x 5 mL |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | dimFormat | Quantity | Price (USD) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
R112022 | Each | 10 x 500 mL | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R060258 | Each | 100 x 2 mL | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R060270 | Each | 100 x 5 mL | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Thermo Scientific™ Remel BHI Broth (Brain Heart Infusion Broth) is for the cultivation of microorganisms.