Double Bagged Bottled Fluid Thioglycollate Medium (FTM)
Thermo Scientific™

Double Bagged Bottled Fluid Thioglycollate Medium (FTM)

Perform confident sterility testing with double bagged bottled media for added security between environmental class changes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFormatCap Type
R112997Dbl Bagged Wide Mouth BottleGold Metal Cap
R112910Dbl Bagged Borosilicate BottlePP plastic cap w/ Teflon
R112647Double Bagged Serum BottleStopper, Crimp seal
Catalog number R112997
Price (CLP)
-
Format:
Dbl Bagged Wide Mouth Bottle
Cap Type:
Gold Metal Cap
Perform confident sterility and microbial limits testing with Thermo Scientific™ Remel™ Double Bagged Bottled Fluid Thioglycollate Medium (FTM) for testing according to USP, JP, and EU regulations and as a general purpose enrichment broth for cultivation and isolation of microorganisms. Double Bagged Bottled Fluid Thioglycollate Medium is ideal for microbiological testing that requires additional packaging for sterility protection between environmental class changes.
Add Remel Double Bagged Bottled Fluid Thioglycollate Medium (FTM) to your sterility and microbial limits testing routine. Our range of validated solutions for the pharmaceutical microbiology laboratory support the production and quality assurance of sterile pharmaceuticals, vaccines, and biotechnology products.

Complete Solutions, Complete Confidence:
  • Secure packaging: double bagged prior to autoclaving to preserve integrity of sterility media
  • Regulatory compliant: USP, JP, and EU harmonized media
  • Adaptable: a range of available fill volumes, media and bottle types to accommodate your sample sizes and performance testing needs
  • Quality guaranteed: Certificates of Analysis with every lot produced
  • Easy-to-use: simply ‘peel and pop’ bags for quick access to the media bottle
  • Multi-purpose media: suitable for general quality testing
Customizable formats and media types are available. Please inquire.

Fluid Thioglycollate Medium (FTM) is a medium for the cultivation of both aerobic and anaerobic organisms in the performance of sterility tests.

This medium is prepared according to the formula specified in the US Pharmacopoeia1 for the performance of sterility tests and conforms to formulations detailed in the British and European Pharmacopoeia.2,3

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
For Laboratory Use
For Laboratory Use

General References:

  1. US Pharmacopoeia. XXIV (2000) Sterility Testing.
  2. Sealey J. Q. (1951) Thesis of the University of Texas.
  3. British Pharmocopoeia. Volume II (2000).
Specifications
Cap TypeGold Metal Cap
DescriptionDouble Bagged Bottled
FormatDbl Bagged Wide Mouth Bottle
Quantity100 mL
Shelf Life273 Days
Product TypeCulture Medium
Unit SizeEach
Contents & Storage
2 to 25°C