Tryptic Soy Broth
Tryptic Soy Broth
Thermo Scientific™

Tryptic Soy Broth

Cultivate and isolate microorganisms with ease using this general purpose enrichment broth with formats available that meet USP guidelines for sterility testing.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityFormatFill Volume
R11888120/Pk.20mm x 113mm Tube15 mL
R065000100/Pk.15mm x 103mm Tube4.5 mL
R07222100/Pk.Tube5 mL
R07226100/Pk.Tube10 mL
R0894820/Pk.20mm x 113mm Tube18 mL
R117832100/Pk.16mm x 125mm Tube9 mL
R11783420/Pk.16mm x 125mm Tube, Hungate Cap10 mL HC
R117835100/Pk.16mm x 125mm Tube, Hungate Cap10 mL HC
R119880100/Pk.20mm x 150mm Tube20 mL
R064890100/Pk.D - 2mL2 mL
R064880100/Pk.D - 1mL1 mL
R0722420/Pk.Tube5 mL
Catalog number R118881
Price (USD)
-
Request A Quote
Quantity:
20/Pk.
Format:
20mm x 113mm Tube
Fill Volume:
15 mL
Cultivate and isolate microorganisms with ease using Thermo Scientific™ Tryptic Soy Broth, a general purpose enrichment broth.
Remove the need for pipetting, dispensing or autoclaving with convenient, pre-filled and ready-to-use TSB tubes.

Part numbers beginning with R11 are for sterility testing according to USP (United States Pharmacopeia).

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Certifications/ComplianceIndustrial Reference: AOAC, BAM, COMPF, SMWW, IDF, SMD, USDA, USP.
DescriptionTryptic Soy Broth
Fill Volume15 mL
Format20mm x 113mm Tube
Quantity20/Pk.
Manufacturing OriginUnited States
Product TypeMicrobiology Media
Unit SizeEach