dITP Solution (100 mM)
Thermo Scientific™

dITP Solution (100 mM)

Thermo Scientific dITP (2'-deoxyinosine 5'-triphosphate) is a highly stable nucleotide supplied as 100 mM aqueous solution titrated to pH 7.3-7.5Read more
Have Questions?
Catalog NumberQuantity
R11910.25 mL
Catalog number R1191
Price (HKD)
720.00
Online Exclusive
Ends: 30-Jun-2026
901.00
Save 181.00 (20%)
Each
Quantity:
0.25 mL
Request bulk or custom format
Price (HKD)
720.00
Online Exclusive
Ends: 30-Jun-2026
901.00
Save 181.00 (20%)
Each

Thermo Scientific dITP (2'-deoxyinosine 5'-triphosphate) is a highly stable nucleotide supplied as 100 mM aqueous solution titrated to pH 7.3-7.5 with NaOH. The nucleotide has greater than 99% purity and is free of nuclease activities and human, and E. coli DNA. Thermo Scientific dITP is designed for use with a wide variety of different molecular biology applications.

Applications
• PCR and RT-PCR with Taq DNA polymerase
• qPCR
• cDNA synthesis
• SNP genotyping
• Primer extension.

Note: dITP is not suitable for PCR with proofreading enzymes or enzyme mixes containing proofreading enzymes.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
PolymeraseDNA Polymerase
Quantity0.25 mL
Shipping ConditionDry Ice
Concentration100 mM
For Use With (Application)Standard PCR, cDNA Synthesis, RT-PCR
FormLiquid
Label or DyeUnlabeled
Unit SizeEach
Contents & Storage
dITP Solution, 100 mM (0.25 mL)

Store at –20°C.