MicroTest™ M4™ Dram, Plastic
MicroTest™ M4™ Dram, Plastic
Thermo Scientific™

MicroTest™ M4™ Dram, Plastic

Save valuable storage space with a glass or plastic smaller container of transport and storage media for viruses, chlamydiae, mycoplasma and ureaplasma.
Have Questions?
Catalog number R12521
Price (USD)
-
Request A Quote
Save valuable laboratory and cold storage space with a significantly smaller container of transport and storage media. The Thermo Scientific™ MicroTest™ M4™ Dram comes in both glass and plastic to meet your needs while maintaining organism viability and counts for verification studies, quality control procedures, and repeat testing of viruses, chlamydiae, mycoplasma and ureaplasma.
Designed for the collection, transportation, maintenance and long-term storage of viral specimens, chlamydiae, mycoplasma and ureaplasma, Thermo Scientific MicroTest products are top quality. From formulation to packaging, MicroTest products maintain the viability of organisms through freeze-thaw cycles while inhibiting antimicrobial contaminants and promoting safety.
 
MicroTest™ M4™ contains gelatin, vancomycin, amphotericin B, and colistin for the transport of viruses, Chlamydiae, Ureaplasmas, and Mycoplasmas.
 
Keep your lab moving in the right direction with a dependable set of solutions for transport, holding, maintenance and long-term culture storage with the MicroTest family of collection and transport media. 
 
Trusted reliable results
  • Stringent quality control, including cellular toxicity on each lot number
  • Patented formulations with stabilizing proteins, buffers, and specific antibiotics
 
Compatibility and viability
  • Provides compatible transport for cell culture, enzyme immunoassays, and molecular diagnostic tests
  • Maintains organism viability and counts for better, more reliable results during validation/verification studies, quality control procedures, and repeat testing
 
Convenient, accommodating format
  • 1.5mL and 3.0mL fill volumes allow multiple tests from one specimen
  • Individual transport media tubes or complete kits offer flexibility and versatility
 
Safe, sturdy packaging
  • Tough, break-resistant tubes with secure, color-coded screw caps prevent leaking and exposure to infectious agents
  • Kits include leak-resistant polypropylene bag for safe sample transport
 
Not all products are available for sale in all territories. Please inquire.
 
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on. 
Specifications
Product TypeDram
Closure ColorBlack
CompatibilityCulture, Shell vial, Freeze-thaw cycles, Culture maintenance, ELISA, PCR, DNA probe technology
DescriptionPlastic Dram
Fill Volume2mL/Dram
For Use With (Application)Viruses: Chlamydiae, Ureaplasmas, and Mycoplasmas
PackagingSingle vial. Packaged in partitioned boxes.
Quantity72 Drams/Pk.
Shelf Life18 Months
Storage Requirements-25°C or 2°C to 8°C
Volume (Metric)2 mL
Unit SizeEach