MicroTest™ Flocked Swab Kits
El producto real puede variar
Thermo Scientific™

MicroTest™ Flocked Swab Kits

Mantenga la viabilidad de los organismos con kits que incluyen medios de transporte de temperatura ambiente de calidad, un hisopo flocado de nailon de punta tradicional y/o un hisopo flocado de nailon de micropunta.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoTipo de puntaTipo de productoIncluye
R12563Tradicional y con micropuntaKit universal
R12565TradicionalKit de hisopos flocadosKit M4RT con hisopo flocado de punta tradicional
R12567Tradicional y con micropuntaKit universalKit universal M4RT con hisopo flocado de micropuntas y punta tradicional
R12561MicrotipKit de hisopos flocadosKit M6 con hisopo flocado de micropuntas
R12569MicrotipKit de hisopos flocadosKit M6 con hisopo flocado de micropuntas
R12560TradicionalKit de hisopos flocadosKit M6 con hisopo flocado de punta tradicional
R12568TradicionalKit de hisopos flocadosKit M6 con hisopo flocado de punta tradicional
R12562Tradicional y con micropuntaKit de hisopos flocadosKit universal M6 con hisopo flocado de micropunta y punta tradicional
R12542MicrotipHisopo flocadoHisopo flocado, micropunta de nailon
Número de catálogo R12563
Precio (MXN)
-
Tipo de punta:
Tradicional y con micropunta
Tipo de producto:
Kit universal

Maintain organism viability and counts for verification studies, quality control procedures, and repeat testing of viruses chlamydiae, mycoplasma and ureaplasma with quality media, proven collection and transport tools, and safe, reliable packaging with Thermo Scientific™ MicroTest™ Flocked Swab Kits.

Designed for the collection, transportation, maintenance and long-term storage of viral specimens, chlamydiae, mycoplasma and ureaplasma, Thermo Scientific MicroTest products are top quality. From formulation to packaging, MicroTest products maintain the viability of organisms through freeze-thaw cycles while inhibiting antimicrobial contaminants and promoting safety.

MicroTest™ M4RT™ contains gelatin, gentamicin, and amphotericin B for the transport of viruses and Chlamydiae.

MicroTest™ M6™ contains gelatin, vancomycin, amphotericin B, and colistin for the transport of viruses, Chlamydiae, Ureaplasmas, and Mycoplasmas.

Microtest Flocked Swab Kits come with 1.5mL or 3mL of liquid medium in various tube types, with or without glass beads, individually wrapped swabs, in either a peel pouch format or zip-seal bag. Please refer to the specific product listing for more information about the kit contents.

Keep your lab moving in the right direction with a dependable set of solutions for transport, holding, maintenance and long-term culture storage with the MicroTest family of collection and transport media.

Trusted reliable results
  • Stringent quality control, including cellular toxicity on each lot number
  • Patented formulations with stabilizing proteins, buffers, and specific antibiotics
Compatibility and viability
  • Provides compatible transport for cell culture, enzyme immunoassays, and molecular diagnostic tests
  • Maintains organism viability and counts for better, more reliable results during validation/verification studies, quality control procedures, and repeat testing
Convenient, accommodating format
  • 1.5mL and 3.0mL fill volumes allow multiple tests from one specimen
  • Individual transport media tubes or complete kits offer flexibility and versatility
Safe, sturdy packaging
  • Tough, break-resistant tubes with secure, color-coded screw caps prevent leaking and exposure to infectious agents
Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Especificaciones
Color del tapónRed
CompatibilidadCultivo, vial Shell, ciclos de descongelación, mantenimiento de cultivo, ELISA, PCR, tecnología de sonda de ADN
DescripciónM6 Universal Kit w/Micro-Tipped and Traditional-Tipped Flocked Swab
Volumen de llenado3 mL
Para utilizar con (aplicación)Virus: Clamidias, ureaplasmas y micoplasmas
Cantidad100 kits/paq.
Shaft MaterialPlástico
Duración de almacenamiento18 meses
Requisitos de almacenamientoDe 2 °C a 30 °C
Tipo de puntaTradicional y con micropunta
Temperatura de transporte2° to 30°C
FormatoTubo
Tipo de mediosM6
Tipo de productoKit universal
Swab ConfigurationDual Applicator (1 Traditional, 1 Microtip Swab)
Unit SizeEach