MicroTest™ Universal Kits
Actual product may vary
MicroTest™ Universal Kits
MicroTest™ Universal Kits
Thermo Scientific™

MicroTest™ Universal Kits

Maintain viability of viruses chlamydiae, mycoplasma and ureaplasma during transport and storage with kits with media and traditional and micro-tipped swabs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Closure Color
R125803 mLGreen
R125703 mLBlue
R125783 mLRed
R125821.5 mLRed
Catalog number R12580
Price (MXN)
-
Volume (Metric):
3 mL
Closure Color:
Green

Maintain organism viability and counts for verification studies, quality control procedures, and repeat testing of viruses, chlamydiae, mycoplasma and ureaplasma with quality media, proven collection and transport tools, and safe, reliable packaging. Thermo Scientific™ MicroTest™ Universal Kits come in four media formulations and include one plastic shaft traditional-tipped polyester swab and one stainless steel shaft micro-tipped polyester swab and a 6 x 9 in. zip-seal bag for safe transport.

Designed for the collection, transportation, maintenance and long-term storage of viral specimens, chlamydiae, mycoplasma and ureaplasma. Thermo Scientific MicroTest products are top quality. From formulation to packaging, MicroTest products maintain the viability of organisms through freeze-thaw cycles while inhibiting antimicrobial contaminants and promoting safety.

MicroTest™ M4™ contains gelatin, vancomycin, amphotericin B, and colistin for the transport of viruses, Chlamydiae, Ureaplasmas, and Mycoplasmas.

MicroTest™ M4RT™ contains gelatin, gentamicin, and amphotericin B for the transport of viruses and Chlamydiae.

MicroTest™ M5™ contains vancomycin, amphotericin B, and colistin and protein stabilizers for the transport of viruses, Chlamydiae, Ureaplasmas, and Mycoplasmas.

MicroTest™ M6™ contains gelatin, vancomycin, amphotericin B, and colistin for the transport of viruses, Chlamydiae, Ureaplasmas, and Mycoplasmas.

MicroTest Universal Kits come with 1.5mL or 3mL of liquid medium in a 15mL conical tube with 3 glass beads, an envelope with one plastic shaft, traditional-tipped polyester swab, one stainless steel shaft micro-tipped polyester swab, and a 6 x 9 in. zip-seal bag.

Keep your lab moving in the right direction with a dependable set of solutions for transport, holding, maintenance and long-term culture storage with the MicroTest family of collection and transport media.

Trusted reliable results

  • Stringent quality control, including cellular toxicity on each lot number
  • Patented formulations with stabilizing proteins, buffers, and specific antibiotics

Compatibility and viability

  • Provides compatible transport for cell culture, enzyme immunoassays, and molecular diagnostic tests
  • Maintains organism viability and counts for better, more reliable results during validation/verification studies, quality control procedures, and repeat testing

Convenient, accommodating format

  • 1.5mL and 3.0mL fill volumes allow multiple tests from one specimen
  • Individual transport media tubes or complete kits offer flexibility and versatility

Safe, sturdy packaging

  • Tough, break-resistant tubes with secure, color-coded screw caps prevent leaking and exposure to infectious agents
  • Kits include leak-resistant polypropylene bag for safe sample transport

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

Specifications
Closure ColorGreen
CompatibilityCulture, Shell vial, Freeze-thaw cycles, Culture maintenance, ELISA, PCR, DNA probe technology (including BDMS, membrane ELISA)
DescriptionM5 Universal Kit
For Use With (Application)Viruses: Chlamydiae, Ureaplasmas, and Mycoplasmas
Quantity100 Kits/Pk.
Shaft MaterialPlastic, Stainless Steel
Shelf Life18 Months
Storage Requirements-25°C or 2°C to 8°C
Tip TypeTraditional and Microtipped
Volume (Metric)3 mL
FormatTube
Product TypeUniversal Kit
Swab ConfigurationDual Applicator (1 Traditional, 1 Microtip Swab)
Unit SizeEach