SDNA Whole Saliva Collection Device
SDNA Whole Saliva Collection Device
SDNA Whole Saliva Collection Device
SDNA Whole Saliva Collection Device
SDNA Whole Saliva Collection Device
Simplify the collection of saliva for DNA/viral RNA biosample collection, transportation, & storage.
Have Questions?
Número de catálogoCantidad
R13100360/caja
Número de catálogo R13100
Precio (MXN)
-
Cantidad:
360/caja
Simplify the collection of saliva for commercial bioinformatic DNA testing applications, academic, research, biobanking, and as a bio-sample collection option for viral DNA/RNA COVID-19 testing for hospital networks, health departments and clinical research organizations with Spectrum™ Solutions SDNA Whole Saliva Collection Device.
The SDNA saliva collection device has been engineered to lead the saliva collection market in molecular diagnostic applications. This self-contained saliva collection system provides critical sample consistency while suspending and neutralizing viral RNA transcripts post collection for sensitive and specific analysis completely inactivating the live virus at ambient temperatures.
  • Single device for DNA/viral RNA biosample collection, transportation, & storage.
  • Intuitive 3-step design, proven to reduce customer collection errors.
  • System maintains biosample consistency.
  • Significantly reduces costs associated with sample failures & recollection.
  • Tube compatible with automated platforms.
  • Innovative, patented chemistry that preserves & protects both DNA/viral RNA transcripts, manages bacteria & mitigates any risk of infection throughout the testing process.
Especificaciones
DescripciónSpectrum Saliva Collection Device
Para utilizar con (aplicación)Commercial bioinformatic DNA testing applications, academic, research, biobanking, and as a biosample collection option for viral DNA/RNA COVID-19 testing
Cantidad360/caja
Unit SizeEach