Yellow Sample Buffer (40X)
Thermo Scientific™

Yellow Sample Buffer (40X)

Thermo Scientific 40X Yellow Sample Buffer is used with Luminaris Color qPCR Master Mixes to color and track the samples.자세히 알아보기
Have Questions?
카탈로그 번호수량
R13814 x 1.25 mL
카탈로그 번호 R1381
제품 가격(KRW)
53,000
온라인 행사
Ends: 31-Dec-2026
66,000
할인액 13,000
Each
수량:
4 x 1.25 mL
제품 가격(KRW)
53,000
온라인 행사
Ends: 31-Dec-2026
66,000
할인액 13,000
Each
Thermo Scientific 40X Yellow Sample Buffer is used with Luminaris Color qPCR Master Mixes to color and track the samples. When using the blue Luminaris Color qPCR Master Mix the qPCR reaction mix is blue before sample addition. After adding the sample the reaction mix turns green, making the overall dispensing of qPCR reaction components quick, easy and error-free. This is particularly important for users of opaque white reaction plates and tubes.

The Yellow Sample Buffer is an aqueous solution of the inert yellow dye (pH 8.0). The buffer is provided as a 40X concentrate and used in 1X concentration in the final reaction (for detailed recommendations refer to the manual of Luminaris Color qPCR Master Mix). The yellow dye does not affect the specificity or sensitivity of qPCR assays. The absorption maximum of the yellow dye is at 413 nm.

Highlights

• Colored sample buffer for easy tracking during pipetting.
For Research Use Only. Not for use in diagnostic procedures.
사양
포함Contains:
• 4 x 1.25 mL 40X Yellow Sample Buffer

Store at -20°C.
제품라인Luminaris
수량4 x 1.25 mL
Unit SizeEach

자주 묻는 질문(FAQ)

Does the yellow dye from the Yellow Sample Buffer provided with Luminaris Color and DyNAmo Flash qPCR Master Mixes influence qPCR performance?

The addition of yellow dye from the Yellow Sample Buffer to qPCR reaction has no influence on the efficiency, specificity, sensitivity or Cq values. Using the Yellow Sample Buffer is optional, but the dye helps to see in which wells the sample has been already added.