NTP Set, Tris buffered
Thermo Scientific™

NTP Set, Tris buffered

Thermo Scientific NTP Set, Tris buffered is a convenient set of 100 mM ATP, CTP, GTP, and UTP solutions. TheseRead more
Have Questions?
Catalog NumberQuantity
R14814 x 0.25 mL
Catalog number R1481
Price (MXN)
-
Quantity:
4 x 0.25 mL
Request bulk or custom format

Thermo Scientific NTP Set, Tris buffered is a convenient set of 100 mM ATP, CTP, GTP, and UTP solutions. These extremely stable nucleotides are supplied as aqueous solutions titrated to pH 7.3-7.5 with Tris base. The nucleotides have greater than 99% purity and are free of nuclease activities and functionally tested in in vitro transcription.

Features
• Purity confirmed by HPLC
• Free of trace contaminating nucleotides, endo-, exodeoxyribonuclease and ribonuclease activities
• Higher in vitro transcription yields with Tris buffered NTPs when compared to NTPs titrated with NaOH
• Stable for 3 years at -20°C, and after multiple freeze-thaw cycles

Applications
• in vitro transcription
• RNA amplification
• siRNA synthesis
• aRNA synthesis

Related products
• ATP, Tris Buffered
• GTP, Tris Buffered
• UTP, Tris Buffered

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)in vitro Transcription, RNA Amplification
FormLiquid
Label or DyeUnlabeled
Quantity4 x 0.25 mL
Shipping ConditionDry Ice
Concentration100 mM
Unit SizeEach
Contents & Storage
NTP Set, Tris buffered, 100 mM (4 x 0.25 mL)

Store at –20°C.