Search
| Número de catálogo | Para utilizar con (equipo) |
|---|---|
| R16001 | 100-240 VAC, 50∼60Hz, 6 Amps |
Maximize efficiency with the first automated culture plate reader. Free up valuable tech time for more meaningful activities with the APAS Independence instrument (Automated Plate Assessment System).
The APAS Independence instrument utilizes the breakthrough Automated Plate Assessment System (APAS) imaging technology and artificial intelligence algorithms to read and categorize agar plates, and remove negatives from the workflow.
Embrace automation in microbiology without the need for large-scale capital resources to streamline with the APAS Independence instrument.
Streamline your workflow
The intuitive nature of the APAS Independence instrument means your workflow remains the same no matter what modules* are used. Training on the Instrument is easy, and module-specific training can be added when required.
Save more than time
Not only does the APAS Independence Instrument save you time by alleviating the bottlenecks in bench reading, it also impacts many aspects of managing an efficient and effective microbiology lab.
Embrace Automation in Microbiology
The APAS Independence is task-focused, compact, and designed specifically to overcome the bottleneck in manual culture plate reading by automatically triaging plates into three categories: 'significant', 'non-significant or 'negative', and 'for review'.
Unlike large-scale automation solutions, the APAS Independence instrument offers the benefits of standardization, but in a small, affordable, and flexible footprint designed to easily integrate with the majority of laboratories.
Availability of product in each country depends on local regulatory marketing authorization status. ©2021 Thermo Fisher Scientific Inc. Clever Culture Systems is the legal manufacturer of the APAS Independence. APAS is a trademark of Clever Culture Systems. All other trademarks are the property of Thermo Fisher Scientific and its subsidiaries. Products are distributed in the U.S. and Europe so uses, applications, and availability of product in each country depend on local regulatory marketing authorization status.