Thermo Scientific™
Remel™ RPR Antigen
Catalog number: R16305-MBD
Thermo Scientific™
Remel™ RPR Antigen
Catalog number: R16305-MBD
Thermo Scientific™ Remel RPR Antigen is used for detection of reagin in human serum.
Catalog Number
R16305-MBD
Unit Size
Each
Quantity
3 Vials/Pk.
Price (USD)
Full specifications
Quantity3 Vials/Pk.
DescriptionRPR Antigen 3 X 3ml Pk./3
Detectable AnalytesReagin
TypeRPR Antigen
Unit SizeEach
Showing 1 of 1
Catalog NumberSpecificationsUnit SizeQuantityPrice (USD)
R16305-MBDFull specifications
Each3 Vials/Pk.Request A Quote
Quantity3 Vials/Pk.
DescriptionRPR Antigen 3 X 3ml Pk./3
Detectable AnalytesReagin
TypeRPR Antigen
Unit SizeEach
Showing 1 of 1
3mL/Vial
Ordering Alerts:
For use with RPR Card Test Kit.