This product is only available through your local Thermo Scientific supplier. Learn where to buy >
Plant gDNA Binding Solution
Catalog number: R1691
Thermo Scientific™  Related applications: DNA Extraction
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
Thermo Scientific GeneJET Plant gDNA Binding Solution is a component of the GeneJET Plant RNA Purification Mini Kit (K0791/K0792) and may be purchased separately. Provided in 40 mL aliquots. Please review protocol manuals to confirm the appropriate solutions for ordering separately.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Buffer Type: Binding Buffer
Product Line: GeneJET
Product Size: 40 mL
Documents
Manuals & protocols