Wash Buffer 2 (concentrated)

Catalog number: R1741

Thermo Scientific™  Related applications: Viral RNA⁄DNA Purification

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Thermo Scientific GeneJET Wash Buffer 2 (concentrated) is a component of several GeneJET nucleic acid purification kits and may be purchased separately. Provided in 11 mL aliquots. Please consult kit protocols to confirm catalog numbers for re-ordering the appropriate kit-specific components.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Buffer Type: Wash Buffer
Product Line: GeneJET
Product Size: 11 ml

Documents

Manuals & protocols