Thermo Scientific™
BactiDisks™ Acinetobacter baumannii ATCC™ 19606™
Catalog number: R19000
Thermo Scientific™
BactiDisks™ Acinetobacter baumannii ATCC™ 19606™
Catalog number: R19000
Thermo Scientific™ BactiDisks™ are lyophilized mixtures of charcoal, gelatin and microorganisms in a stable disk format.
Catalog Number
R19000
Unit Size
-
Quantity
10 Disk/Vial
Price (USD)
Full specifications
ATCC®19606™*
DescriptionAcinetobacter baumannii ATCC™ 19606™
FormatBactiDisks
Product TypeBactidisk
Target Organism ClassGram-Negative Rods
Test TypeQualitative QC
Quantity10 Disk/Vial
Unit Size-
Showing 1 of 1
Catalog NumberSpecificationsUnit SizeQuantityPrice (USD)
R19000Full specifications
-10 Disk/VialRequest A Quote
ATCC®19606™*
DescriptionAcinetobacter baumannii ATCC™ 19606™
FormatBactiDisks
Product TypeBactidisk
Target Organism ClassGram-Negative Rods
Test TypeQualitative QC
Quantity10 Disk/Vial
Unit Size-
Showing 1 of 1
The disk is dissolved in 0.1mL of appropriate culture broth, inoculated onto an appropriate culture medium, and incubated at the appropriate temperature, atmosphere and time. BactiDisks are recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures. Packaged 10 disks per vial.
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Figures