Informational:This product is unavailable. For information on equivalent strain with Thermo Scientific™ Culti-Loops™ product portfolio, please contact your local Thermo Scientific representative or customer service department.
Thermo Scientific™
BactiDisks™ Aggregatibacter aphrophilus ATCC™ 7901™
Catalog number: R19035
Thermo Scientific™
BactiDisks™ Aggregatibacter aphrophilus ATCC™ 7901™
Catalog number: R19035
Thermo Scientific™ BactiDisks™ are lyophilized mixtures of charcoal, gelatin and microorganisms in a stable disk format.
Catalog Number
R19035
Unit Size
Each
Description
Aggregatibacter aphrophilus
Price (USD)
Full specifications
ATCC®7901™*
DescriptionAggregatibacter aphrophilus
FormatBactiDisks
Product TypeBactidisk
Target Organism ClassFastidious
Test TypeQualitative QC
Type10 Disks/Vl.
Quantity10 Disk/Vial
Strain Designations430 [ICPB 3506]
Unit SizeEach
Showing 1 of 1
Catalog NumberSpecificationsUnit SizeDescriptionPrice (USD)
R19035Full specifications
EachAggregatibacter aphrophilusRequest A Quote
ATCC®7901™*
DescriptionAggregatibacter aphrophilus
FormatBactiDisks
Product TypeBactidisk
Target Organism ClassFastidious
Test TypeQualitative QC
Type10 Disks/Vl.
Quantity10 Disk/Vial
Strain Designations430 [ICPB 3506]
Unit SizeEach
Showing 1 of 1
The disk is dissolved in 0.1mL of appropriate culture broth, inoculated onto an appropriate culture medium, and incubated at the appropriate temperature, atmosphere and time. BactiDisks are recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures. Packaged 10 disks per vial.
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.