Thermo Scientific™
BactiDisks™ Staphylococcus epidermidis ATCC™ 12228™
 Related applications:
Clinical Microbiology
Thermo Scientific™ BactiDisks™ are lyophilized mixtures of charcoal, gelatin and microorganisms in a stable disk format.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
The disk is dissolved in 0.1mL of appropriate culture broth, inoculated onto an appropriate culture medium, and incubated at the appropriate temperature, atmosphere and time. BactiDisks are recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures. Packaged 10 disks per vial.
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Documents
Manuals & protocols