BactiDisks™ Streptococcus sp. Type 4 group F ATCC™ 9328™
This product is unavailable. For information on equivalent strain with Thermo Scientific™ Culti-Loops™ product portfolio, please contact your local Thermo Scientific representative or customer service department.
BactiDisks™ Streptococcus sp. Type 4 group F ATCC™ 9328™
Thermo Scientific™
BactiDisks™ Streptococcus sp. Type 4 group F ATCC™ 9328™
Thermo Scientific™ BactiDisks™ are lyophilized mixtures of charcoal, gelatin and microorganisms in a stable disk format.
Have Questions?
Catalog number R19177
Price (USD)/ Each
-
Request A Quote
Quantity:
10 Disk/Vial
Thermo Scientific™ BactiDisks™ are lyophilized mixtures of charcoal, gelatin and microorganisms in a stable disk format.
The disk is dissolved in 0.1mL of appropriate culture broth, inoculated onto an appropriate culture medium, and incubated at the appropriate temperature, atmosphere and time. BactiDisks are recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures. Packaged 10 disks per vial.
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Specifications
ATCC®9328TM*
DescriptionStreptococcus sp. Type 4 group F ATCC™ 9328™
FormatBactiDisks
Product TypeBactidisk
Target Organism ClassCocci
Test TypeQualitative QC
Quantity10 Disk/Vial
Unit SizeEach