BactiDisks™ Streptococcus bovis ATCC™ 35034™
This product is unavailable. For information on equivalent strain with Thermo Scientific™ Culti-Loops™ product portfolio, please contact your local Thermo Scientific representative or customer service department.
BactiDisks™ Streptococcus bovis ATCC™ 35034™
Thermo Scientific™
BactiDisks™ Streptococcus bovis ATCC™ 35034™
Thermo Scientific™ BactiDisks™ are lyophilized mixtures of charcoal, gelatin and microorganisms in a stable disk format.
Have Questions?
Catalog number R19178
Price (USD)/ Each
-
Request A Quote
Quantity:
10 Disk/Vial
Thermo Scientific™ BactiDisks™ are lyophilized mixtures of charcoal, gelatin and microorganisms in a stable disk format.
The disk is dissolved in 0.1mL of appropriate culture broth, inoculated onto an appropriate culture medium, and incubated at the appropriate temperature, atmosphere and time. BactiDisks are recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures. Packaged 10 disks per vial.
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Specifications
ATCC®35034TM*
DescriptionStreptococcus bovis ATCC™ 35034™ Group D
FormatBactiDisks
Product TypeQuality Control
Target Organism ClassCocci
Test TypeQualitative QC
Quantity10 Disk/Vial
Strain DesignationsWDCM00058
Unit SizeEach