Search Thermo Fisher Scientific
ATCC® | 33314TM* |
Description | Pediococcus pentosaceus ATCC™ 33314™ |
Product Type | Quality Control |
Target Organism Class | Anaerobes |
Test Type | Qualitative QC |
Quantity | 10 Disk/Vial |
Strain Designations | NCDO 1859 [DSM 20333, JCM 5885] |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Quantity | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
R19355 | Each | 10 Disk/Vial | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
The disk is dissolved in 0.1mL of appropriate culture broth, inoculated onto an appropriate culture medium, and incubated at the appropriate temperature, atmosphere and time. BactiDisks are recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures. Packaged 10 disks per vial.