Remel™ McFarland Equivalence Turbidity Standards
Remel™ McFarland Equivalence Turbidity Standards
Thermo Scientific™
Remel™ McFarland Equivalence Turbidity Standards
Thermo Scientific™ Remel McFarland Equivalence Turbidity Standard is packaged in plastic case with a visual comparison card.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
R20413Turbidity Standard 3.0
R20421Turbidity Standard Set (1 each of 0.5, 1.0, 2.0, 3.0, 4.0)
R20414Turbidity Standard 4.0
R20412Turbidity Standard 2.0
R20415Turbidity Standard 5.0
R20410Turbidity Standard 0.5
R20411Turbidity Standard 1.0
Catalog number R20413
Price (USD)
-
Request A Quote
Description:
Turbidity Standard 3.0
Thermo Scientific™ Remel McFarland Equivalence Turbidity Standard is packaged in plastic case with a visual comparison card.
  • Tube size is 15 x 103mm and fits in most spectrophotometers
  • Polystyrene technology allows longer shelf life
Specifications
DescriptionTurbidity Standard 3.0
Tubing Size15 x 103 mm
Unit SizeEach