PYR/Esculin Disc
Thermo Scientific™
PYR/Esculin Disc
Presumptively identify Group A streptococci and enterococci using PYR/Esculin Disk.
Have Questions?
Número de catálogoCantidad
R2113825 discos/vial
Número de catálogo R21138
Precio (MXN)
-
Cantidad:
25 discos/vial
Rapidly, presumptively identify Group A streptococci and enterococci using reagent-impregnated Thermo Scientific™ Remel™ PYR/Esculin Disk. Ellner et al. described a colorimetric method for the PYR test using filter paper strips containing PYR substrate1. Using a rapid spot test, Edberg et al. demonstrated the hydrolysis of esculin to esculetin by utilizing the loss of fluorescence as an indicator of esculin hydrolysis2.
In 1981, Godsey et al. described a test to differentiate group A streptococci and enterococci from other streptococci based on their ability to cleave L-pyrrolidonyl-β-naphthylamide (PYR)3. In 1982, Facklam et al. used PYR test in conjunction with the CAMP and esculin hydrolysis tests to presumptively identify streptococci4,5.
  • Ready to use - L-pyrrolidonyl-β-naphthylamide-impregnated disk
  • Easy to interpret - With visual inspection
The enzyme pyrrolidonyl peptidase hydrolyzes the substrate L-pyrrolidonyl-β-naphthylamide (PYR) to produce β-naphthylamine. Following addition of N,N-dimethyl-aminocinnamaldehyde (PYR reagent) a red color is formed. Escuin is hydrolyzed at the β- glucose linkage to yield two products – esculetin and glucose. Esculin fluoresces under ultraviolet light at 360 nm, whereas the hydrolysis product esculetin fails to fluoresce. Both reactive substrates, PYR and esculin, are lyophilized in disk form for stability.
  1. Ellner, P.D., D.A. Williams, M.E. Hosmer, and M.A. Cohenford. 1985. J. Clin. Microbiol. 22:880-881.
  2. Edberg, S.C., K. Gam, C.J. Bottenbley , and J.M. Singer. 1976. J. Clin. Microbiol. 4:180-184.
  3. Godsey, J., R. Schulman, and L. Eriquez. 1981. Abstract C84:276. Abstratcs of the 81st General Meeting of the American Society for Microbiology. ASM, Washington , D.C.
  4. Fackalm, R.R., L.G. Thacker, B. Fox, L. Eriquez. 1982. J. Clin. Microbiol. 15:987-990.
  5. Christie, R., N.E. Atkins, E. Munch-Petersen. 1944. Aust. J. Exp. Biol. Med. Sci. 22:197-200.
Especificaciones
DescripciónPYR/Esculin Disk
FormatoVial
Cantidad25 discos/vial
Unit SizeEach
Contenido y almacenamiento
De 2 °C a 8 °C