SPS Disk
Thermo Scientific™
SPS Disk
SPS Disk is a reagent-impregnated disk recommended for presumptive identification of Peptostreptococcus anaerobius.
Have Questions?
Catalog NumberQuantity
R2114025 Disks/Vial
Catalog number R21140
Price (USD)
-
Quantity:
25 Disks/Vial
Presumptively identify Peptostreptococcus anaerobius using Thermo Scientific™ Remel™ SPS Disk. Graves et al. used a reagent-impregnated disk to determine the susceptibility of anaerobic cocci to SPS. They reported that amongst anaerobic cocci only P. anaerobius is sensitive to SPS. Wideman and Wilkins further verified these findings and reported 98% overall accuracy in the identification of P. anaerobius isolated from clinical specimens1,2.
Peptostreptococcus anaerobius is a slow growing, Gram-positive anaerobic cocci (GPAC) bacterium. P. anaerobius is an opportunistic pathogen that can cause multiple deep organ abscesses and infections throughout the body, especially in the abdominal cavity and female urogenitary tract.
Use SPS Disk for presumptive identification of Peptostreptococcus anaerobius.
  • Ready to use - Reagent impregnated disk
  • Easy to store - No special storage required
  • Easy to interpret - Visible zone of inhibition
    • Sensitive - Zone of inhibition equal to or greater than 12 mm
    • Resistant - No zone of inhibition or zone less than 12 mm
SPS Disk contains sodium polyanethol sulfonate. Sodium polyanethol sulfonate (SPS) is a synthetic, heat stable anticoagulant which inhibits bactericidal agents in blood3. SPS has also been found to inhibit the growth of certain anaerobic cocci. The growth of P. anaerobius is inhibited by SPS. Positive test (sensitive) is indicated by a zone diameter of 12-27 mm around SPS disk.
  1. Wideman, P.A., V.L. Vargo, D. Citronbaum, and S.M. Finegold. 1976. J.Clin.Microbiol. 4:330-333.
  2. Wilkins, T.D. and S.E.H. West. 1976. J.Clin.Microbiol. 3:393-396.
  3. Graves, M.H., J.A. Morello, and F.E. Kocka. 1974. Appl.Microbiol. 27:1131-1133.
Specifications
DescriptionSPS Disk
FormatReagent Disk
Quantity25 Disks/Vial
Detectable AnalytesP. anaerobius
Unit SizeEach
Contents & Storage
2°C to 8°C