Product Image
Thermo Scientific™

Remel™ BactiDrop™ Ehrlich's Indole

Remel BactiDrop Ehrlich's Indole is used for detection of the tryptophan enzyme.
Have Questions?
Catalog NumberQuantity
R2152050 x 0.75 mL
Catalog number R21520
Price (HKD)
-
Quantity:
50 x 0.75 mL

Remel BactiDrop Ehrlich's Indole is used for detection of the tryptophan enzyme.

Recommended for use with nonfermenters and anaerobes grown in Indole Broth.

Ordering Alerts:

BactiDrop reagents are supplied in crushable ampules encased in a protective plastic sheath with a convenient dropper dispenser tip.

Specifications
DescriptionBactiDrop Indole, Ehrlich's
Quantity50 x 0.75 mL
CE MarkerYes
Detectable AnalytesSkatole
Unit SizeEach
Contents & Storage
20°C to 25°C