Product Image
Thermo Scientific™

Remel™ BactiDrop™ Voges-Proskauer A

Determine the ability of gram-negative bacilli to produce acetylmethylcarbinol (acetoin) from glucose fermentation using Voges-Proskauer A Test.
Have Questions?
Número de catálogoCantidad
R2156050 x 0,75 mL
Número de catálogo R21560
Precio (MXN)
-
Cantidad:
50 x 0,75 mL
Determine the ability of gram-negative bacilli to produce acetylmethylcarbinol (acetoin) from glucose fermentation using Thermo Scientific™ Remel™ Bactidrop™ Voges-Proskauer A Test. In 1898, Voges and Proskauer reported production of acetylmethylcarbinol from glucose fermentation by certain bacteria when grown in an appropriate medium such as MR-VP Broth, which is a modification of Clark and Lubs medium1,2. Later Barret made the VP test more sensitive by using 5% Alpha Naphthol (VP-A) to the medium for the detection of acetylmethylcarbinol3.
The BactiDrop™ Voges-Proskauer A test is based on the detection of acetoin (acetylmethylcarbinol) production which is a precursor to 2,3-butanediol from glucose metabolism.Ready to use: Convenience of a prepared reagent.Ease of Differentiation: VP positive organisms produce a pink color development while VP negative organisms produce no color.Rapid Detection: Within 15 minutes.Glucose is metabolized to pyruvic acid from which the neutral products acetoin and 2.3-butanediol are formed. One molecule of acetoin is formed by the decarboxylation of two molecules of pyruvic acid. In the presence of oxygen and alkali, acetoin and 2,3-butanediol are oxidized to diacetyl which is the reactant for the pink color produced in Voges-Proskauer test. 5% Alpha Naphthol is a catalyst which acts as a color intensifier and increase the sensitivity of the reaction.Used in conjunction with VP-B, Part No. R21562. For use with MRVP Broth.Not all products are available for sale in all territories. Please inquire.Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
  1. Voges O. and Proskauer B., 1898, Z. Hyg. 28:20-32. Clark W. M. and H.A. Lubs, 1915, J. Infect. Dis., 17:160-173.
  2. Barritt, M.M. 1936. J. Pathol. Bacteriol. 42:441-454.
Especificaciones
DescripciónBactidrop Vp-a Alpha Na
Cantidad50 x 0,75 mL
Marcado CESí
Analitos detectablesAcetylmethylcarbinol
Unit SizeEach
Contenido y almacenamiento
De 20 °C a 25 °C