Product Image
Thermo Scientific™

Remel™ BactiDrop™ Voges-Proskauer B

Determine the ability of gram-negative bacilli to produce acetylmethylcarbinol (acetoin) from glucose fermentation using Voges-Proskauer B Test.
Have Questions?
Catalog NumberQuantity
R2156250 x 0.75 mL
Catalog number R21562
Price (USD)
-
Quantity:
50 x 0.75 mL
Determine the ability of gram-negative bacilli to produce acetylmethylcarbinol (acetoin) from glucose fermentation using Thermo Scientific™ Remel™ Bactidrop™ Voges-Proskauer B Test. In 1898, Voges and Proskauer reported production of acetylmethylcarbinol from glucose fermentation by certain bacteria when grown in an appropriate medium such as MR-VP Broth which is a modification of Clark and Lubs medium 1,2. Later Barret made the VP test more sensitive by using 40% Potassium Hydroxide (VP-B) to the medium for the detection of acetylmethylcarbinol production3.
The BactiDrop™ Voges-Proskauer test is based on the detection of acetylmethylcarbinol production which is a precursor to 2,3-butanediol from glucose metabolism.Ready to use: Convenience of a prepared reagent.Ease of differentiation: VP positive organisms produce a pink color development while VP negative organisms produce no color.Rapid detection: Within 15 minutes.Glucose is metabolized to pyruvic acid from which the neutral products acetoin and 2.3-butanediol are formed. One molecule of acetoin is formed by the decarboxylation of two molecules of pyruvic acid. In the presence of oxygen and alkali, acetoin and 2,3-butanediol are oxidized to diacetyl which is the reactant for the pink color produced in Voges-Proskauer test. 40% potassium hydroxide aids in the absorption of CO2 to hasten the oxidation of acetoin to diacetyl.Used in conjunction with VP-A, Part No. R21560. For use with MRVP Broth.Not all products are available for sale in all territories. Please inquire.Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
  1. Voges O. and Proskauer B., 1898, Z. Hyg. 28:20-32.
  2. Clark W. M. and H.A. Lubs, 1915, J. Infect. Dis., 17:160-173.
  3. Barritt, M.M. 1936. J. Pathol. Bacteriol. 42:441-454.
Specifications
DescriptionBactiDrop Voges-Proskauer B
Quantity50 x 0.75 mL
CE MarkerYes
Detectable AnalytesAcetylmethylcarbinol
Unit SizeEach
Contents & Storage
20°C to 25°C