Elution Buffer for EpiJET Bisulfite Conversion Kit
Catalog number: R2161
Thermo Scientific™  Related applications: Methylation Analysis
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
Thermo Scientific EpiJet Elution Buffer is a component of the EpiJET Bisulfite Conversion Kit and may be purchased separately. Provided in 1.5 mL aliquots.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Buffer Type: Other Buffers
Product Line: EpiJET
Product Size: 1.5 mL
Documents
Manuals & protocols