Product Image
Thermo Scientific™

Remel™ Fecal Transport Two-Vial Sets: Cary-Blair w/Indicator/Empty Vial

Remel Fecal Transport Two-Vial Sets: Cary-Blair w/Indicator/Empty Vial offer preservation and recovery of enteric bacterial pathogens.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoVolumen (métrico)
R2194930 mL
R2161615 mL
Número de catálogo R21949
Precio (MXN)
-
Volumen (métrico):
30 mL

Remel Fecal Transport Two-Vial Sets: Cary-Blair w/Indicator/Empty Vial offer preservation and recovery of enteric bacterial pathogens.

Caps are color coded to match industry standard colors and have an attached spork.
  • Broad range with long shelf life
  • Industry standard color-coded caps and labels
  • Easy to use spork cap for safe collection
  • Convenient packaging, including bulk kits
  • Multilingual patient collection instructions
  • Empty Vial is used for test procedures not compatible with fecal preservatives
  • Cary-Blair with Indicator contains 0.16% agar and is for preservation and recovery of enteric bacterial pathogens
Especificaciones
DescripciónCary-Blair/Empty Vial; Two-vial Sets
Envase2-Vial Set
Volumen (imperial)1.01 oz.
Volumen (métrico)30 mL
Tipo de productoFecal Transport Set
Unit SizeEach